Skip to Content
Merck
All Photos(2)

Documents

16866

Supelco

Monoethylglycinexylidine

analytical standard

Synonym(s):

MEGX, MGEX, Monoethylglycinexylidide, N-(2,6-dimethylphenyl)-2-(ethylamino)acetamide, Norlidocaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O
CAS Number:
Molecular Weight:
206.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

SMILES string

CC1=C(NC(CNCC)=O)C(C)=CC=C1

InChI

1S/C12H18N2O/c1-4-13-8-11(15)14-12-9(2)6-5-7-10(12)3/h5-7,13H,4,8H2,1-3H3,(H,14,15)

InChI key

WRMRXPASUROZGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Pharmacokinetic Analysis: A study by Liu ZY et al. examines the pharmacokinetics of intravenous and topical lidocaine in patients undergoing thoracoscopic pulmonary resection. This research, which could integrate compounds like Monoethylglycinexylidine for analytical validation, highlights the importance of understanding drug dynamics for improving surgical outcomes (Liu et al., 2024).

  • Veterinary Pharmacology: Knych HK et al. explore the pharmacokinetics and metabolism of lidocaine in horses following a palmar digital nerve block. Studies like this could benefit from the use of Monoethylglycinexylidine as a standard for accurate drug measurement and monitoring in veterinary medicine, ensuring safe and effective dosing (Knych et al., 2023).

  • Anesthesia Research: Crouch CE et al. provide insights into the pharmacokinetics of lidocaine intraoperative infusion during partial hepatectomy for living liver donation. Research utilizing Monoethylglycinexylidine could enhance the understanding of local anesthetics′ systemic effects, contributing to safer anesthetic practices (Crouch et al., 2023).

  • Anesthetic Efficacy in Avian Species: Escobar A et al. investigate the effects of two continuous infusion doses of lidocaine on isoflurane minimum anesthetic concentration in chickens. Incorporating Monoethylglycinexylidine in such studies could refine our understanding of dose-dependent effects and optimize anesthetic protocols for non-mammalian species (Escobar et al., 2023).

  • Clinical Pharmacokinetics: Jin Y et al. discuss the simultaneous determination of lidocaine and its active metabolites in plasma by UPLC-MS/MS, applied to a clinical pharmacokinetic study in liver cancer patients. Monoethylglycinexylidine could be used to validate and calibrate assays, enhancing the reliability of pharmacokinetic data in clinical settings (Jin et al., 2022).

Biochem/physiol Actions

Monoethylglycinexylidide (MEGX) is an active metabolite of lidocaine.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service