Skip to Content
Merck
All Photos(1)

Documents

04004

Sigma-Aldrich

Phosphorus, red

≥97.0%

Synonym(s):

P

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
P
CAS Number:
Molecular Weight:
30.97
EC Number:
MDL number:
UNSPSC Code:
12141909
PubChem Substance ID:
NACRES:
NA.22

vapor density

0.02 (vs air)

Quality Level

vapor pressure

0.03 mmHg ( 21 °C)
1 mmHg ( 76.6 °C)

Assay

≥97.0%

autoignition temp.

500 °F

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

resistivity

10 μΩ-cm, 20°C

mp

416 °C (subl.) (lit.)

density

2.34 g/mL at 25 °C (lit.)

cation traces

Fe: ≤2000 mg/kg

storage temp.

room temp

SMILES string

[P]

InChI

1S/P

InChI key

OAICVXFJPJFONN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Red phosphorus is the most common allotrope ofphosphorus due to its relative stability compared to white phosphorus. It has a variety of uses including, but notlimited to, use as a catalyst, battery anode, flame retardant, and igniter.

Application

Red phosphorus can be used as a:
  • Photocatalyst for the formation of H2 from water.
  • Catalyst in the synthesis of tris(2-pyridyl)phosphine by reacting with 2-bromopyridine in the presence of a base.
  • Flame retardant along with aluminum oxide in recycled poly(ethylene terephthalate).
  • Anode material in combination with carbon composites for sodium-ion batteries.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An amorphous red phosphorus/carbon composite as a promising anode material for sodium ion batteries
Kim Y, et al.
Advanced Materials, 25(22), 3045-3049 (2013)
Expedient one-pot organometallics-free synthesis of tris (2-pyridyl) phosphine from 2-bromopyridine and elemental phosphorus
Trofimov BA, et al.
Tetrahedron Letters, 53(19), 2424-2427 (2012)
Red phosphorus/aluminium oxide compositions as flame retardants in recycled poly (ethylene terephthalate
Laoutid F, et al.
Polymer Degradation and Stability, 82(2), 357-363 (2003)
Special materials in pyrotechnics: V. Military applications of phosphorus and its compounds
Koch E-C
Propellants, Explosives, Pyrotechnics, 33(3), 165-176 (2008)
New Evidence Concerning the Structure of Amorphous Red Phosphorus
Hartl H
Angewandte Chemie (International Edition in English), 34(23-24), 2637-2638 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service