Skip to Content
Merck
All Photos(3)

Key Documents

NXF-50

Millipore

Natrix® Q Process 600

Synonym(s):

Natrix® Q Membrane Adsorber

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32011308

ligand

quaternary amine

Quality Level

product line

Natrix®

parameter

2.3-11.5 L/min flow rate

bed H

0.5 mm

bed volume

460 mL

matrix active group

Macroporous polymer

pore size

0.4 μm pore size

capacity

92 g binding capacity (BSA)(per device)

separation technique

strong anion exchange

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Strong anion exchange chromatography membrane design to purify biomolecules with a high density of quaternary amine ligand the enables fast flow rates and high throughput at high loads while maintaining excellent impurity reduction
With a revolutionary three-dimensional macroporous hydrogel structure that provides a high density of binding sites and rapid mass transfer, Natrix® chromatography membranes deliver binding capacity that exceeds resin-based columns with fast flow rates typical of traditional membrane adsorbers. This combination of performance and speed in a single-use, plug-and-play device format enables low-risk, scalable solutions for efficient biomolecule purification

Natrix® Q chromatography membrane is a high productivity strong anion exchange membrane designed for biomolecule purification. The unique Natrix® Q chromatography membrane is a porous polyacrylamide hydrogel containing a high density of pendant quaternary ammonium (Q) binding groups that are directly polymerized within the an inert, macroporous membrane scaffold. The interconnected pore structure and high functional group density enables fast flow rates (seconds of residence time) with impressive throughput to extremely high loads while maintaining excellent impurity reduction across a wide range of buffer conditions. The Natrix Q chromatography membrane design delivers improvements in productivity, flexibility, and process robustness and economics for any bioprocess design. The Natrix® Q Process 600 is process scale device designed for full-scale clinical and commercial manufacturing.

mAb nominal polishing capacity: 4600 g

Membrane configuration: Pleated

1 device per package

Features and Benefits

Natrix® Q membrane adsorber provides users with:

  • High density of quaternary amine binding sites enabling rapid mass transfer
  • High binding capacity even at fast flow rates
  • Combination of performance and speed in, single use, plug-and-play device format enables low risk, scalable solutions for efficient biomolecule purification.

Packaging

Double bagged

Legal Information

6538 is a trademark of American Type Culture Collection
NATRIX is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mochao Zhao et al.
Vaccine, 37(36), 5491-5503 (2018-04-09)
Affinity chromatography is among the most powerful separation techniques, achieving the finest separation with high yields even in the most challenging feed streams. Incorporating affinity chromatography in vaccine purification has long been attempted by researchers to improve unit yield and
Renaud Jacquemart et al.
Computational and structural biotechnology journal, 14, 309-318 (2016-08-30)
The current processing paradigm of large manufacturing facilities dedicated to single product production is no longer an effective approach for best manufacturing practices. Increasing competition for new indications and the launch of biosimilars for the monoclonal antibody market have put
Ying Hou et al.
Biotechnology progress, 31(4), 974-982 (2015-05-29)
Protein A chromatography is widely employed for the capture and purification of monoclonal antibodies (mAbs). Because of the high cost of protein A resins, there is a significant economic driving force to seek new downstream processing strategies. Membrane chromatography has

Articles

Membrane adsorbers are a cost-effective, robust, single-use solution for monoclonal antibody polishing

See case study examples of how to optimize chromatographic purification of plasmid DNA for Biopharmaceutical Applications.

See case study examples of how to optimize chromatographic purification of plasmid DNA for Biopharmaceutical Applications.

See case study examples of how to optimize chromatographic purification of plasmid DNA for Biopharmaceutical Applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service