Skip to Content
Merck
All Photos(1)

Key Documents

NM0044

Millipore

NovaSeal Battery Powered Crimping Tool

AC/DC input 220 V / 240 V AC, EuroPlug, Kit content : crimping tool, 10,8V Li-ion battery, battery charger

Synonym(s):

NovaSeal Electrical Crimping Tool

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
eCl@ss:
32010310

Quality Level

sterility

non-sterile

product line

NovaSeal

compatibility

for use with NovaSeal pinch pipes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service