Skip to Content
Merck
All Photos(3)

Key Documents

MZAAWG101

Millipore

Microfil® with EZ-Pak®

sample volume 100 mL, mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, white filter, gridded filter

Synonym(s):

Membrane filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24

material

gridded filter
mixed cellulose esters (MCE)
polypropylene
white filter

Agency

ISO 7704

description

Yeast or mold

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic

manufacturer/tradename

Microfil® with EZ-Pak®

parameter

100 mL sample volume

filter diam.

47 mm

funnel diam.

100 mL

matrix

MF-Millipore

pore size

0.8 μm pore size

input

sample type food(s)
sample type cider
sample type wine
sample type soft drink
sample type beer
sample type beverage(s)
sample(s) (environmental)
sample type cosmetics
sample type sport drink
sample type bottled water

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

For the ultimate in convenience and efficiency, combine the EZ-Pak system with the Microfil filtration system. The Microfil system includes ready-to-use, sterile filtration funnels and a unique filtration support to facilitate sample processing. No preparation steps are required.
The Microfil filtration system is a simple and reliable solution for routine microbiological testing. These ready-to-use, 100 mL Microfil funnels that comes with the 0.8 μm pore size, white gridded EZ pak filter system eliminate the need for washing and sterilizing after each test thus making the equipment handling easier and efficient to retain and recover microorganism like yeast and molds. The push-fit design of the Microfil funnels seals tightly to manifold supports without the need for clamps or O-rings. Unlike with conventional filter holders, incomplete sealing at the membrane/funnel interface is eliminated resulting in consistent recovery of organisms.

Application

These filters find application in Clinical, Cosmetics, Environmental, Food and Beverage, and Industrial QC. Microbial analysis of beer, bottled water, cider, sport drink and wine by membrane filtration method
Yeast or mold

Principle

Vacuum

Physical form

Color Code: White

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service