Skip to Content
Merck
All Photos(2)

Key Documents

MXQUA4K01

Millipore

Milliflex® Quantum Standard Kit

with validation protocol A4 format, For the rapid detection of microbial contamination

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.79

manufacturer/tradename

Milliflex®

input

sample(s) (raw material)
water
sample(s) (in process)
sample(s) (environmental)
sample(s) (final product)

application(s)

pharmaceutical

shipped in

ambient

General description

Milliflex® Quantum System is a rapid fluorescence-based technology for fast quantitative detection of contaminants over a broad range of filterable matrices. This easy-to-use system detects viable microorganisms filtered on a membrane down to one CFU per sample. The non-destructive method allows the identification, by any method, of any microorganisms detected during the initial fluorescence reading. The sample is prepared by membrane filtration using the Milliflex® system to ensure consistent and reliable results. The kit is also provided with a validation protocol for faster and easier validation and ensuring system′s performance is compliant with its validated specifications.

Application

The Milliflex® Quantum system is used for the rapid detection of microbial contamination in raw materials, in-process samples, final products, water & environmental samples.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service