Skip to Content
Merck
All Photos(1)

Key Documents

MSGVN2B50

Millipore

Multiscreen® 96 well Plate, hydrophilic PVDF membrane

pore size 0.22 μm, non-sterile

Synonym(s):

96-Well Hydrophilic Plate, Hydrophilic 96-Well Plate, PVDF 96-Well Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

material

Barex®
PVDF membrane (hydrophilic)
flat bottom wells
titanium dioxide

Quality Level

description

Receptor binding assays on whole cells and cell fragments, protein kinase/phosphatase precipitation assays, bead/resin based assays

sterility

non-sterile

product line

MultiScreen®

feature

hydrophilic
lid

manufacturer/tradename

MultiScreen®

parameter

50-250 μL sample volume (per well)

technique(s)

activity assay: suitable (protein kinase/phosphatase precipitation)
bead-based assay: suitable
ligand binding assay: suitable
receptor binding assay: suitable

H

14.4 mm

L

128 mm

W

85.5 mm

filtration area

0.28 cm2

plate size

96 wells

well maximum volume

300 μL

working volume

50-250 μL

matrix

Durapore®

pore size

0.22 μm pore size

binding type

low binding surface

shipped in

ambient

General description

MultiScreen® HTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.

Application

MultiScreen® HTS GV Filter Plate, 0.22 μm, opaque, non-sterile has been used in luciferase immunoprecipitation systems (LIPS) assay

Legal Information

Barex is a registered trademark of INEOS Barex AG
Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Whitehouse et al.
Vaccine, 34(15), 1792-1799 (2016-02-28)
Staphylococcus aureus is a pathogen which causes life-threatening infection, the incidence of which rises during adult life. This, together with the emergence of drug-resistant strains and the expansion of more susceptible elderly populations, represents the rationale for the ongoing development

Articles

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

Related Content

This provides information about the chemical compatibility of MultiScreen Filter Plates with different reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service