Skip to Content
Merck
All Photos(2)

Key Documents

MISP00011

Millipore

Microfil® O-ring Kit

For 3 valves and use with Microfil® filtration systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

Quality Level

packaging

pkg of 3 sets

manufacturer/tradename

Microfil®

input

sample type beverage(s)
sample type food(s)

application(s)

bioburden testing
clinical
environmental
food and beverages
industrial qc
water monitoring

compatibility

for use with Microfil® filtration system

shipped in

ambient

General description

The O-ring Kit for 3 valves is an additional spare part of the Microfil® filtration system. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. The O-rings help in tight sealing of the funnel to the Microfil® thereby, aiding in the leak-free operation and uniform recovery of microorganisms.

Application

The O-ring Kit for 3 valves is an additional accessory of the Microfil® filtration system used in general microbiological analysis of beer, bottled water, pharmaceutical products analysis, environmental analysis, bioburden testing, and water monitoring.

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service