Skip to Content
Merck
All Photos(1)

Key Documents

MAB1976F

Sigma-Aldrich

Anti-Integrin αVβ3 Antibody, clone LM609, FITC conjugated

clone LM609, Chemicon®, from mouse

Synonym(s):

Vitronectin Receptor, CD51/CD61

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

LM609, monoclonal

species reactivity

canine, human, monkey, rabbit, pig, bovine, avian, chicken

should not react with

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG1

suitability

not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

General description

The integrin family of cell adhesion receptors consists of at least 16 membrane-associated heterodimers, composed of an alpha and beta subunit that associate in a non-covalent manner. The structure and functional diversity of the integrin family are based upon the pairing abilities of the individual alpha and beta subunits. Key to these molecular interactions between the integrin receptors and their respective ligands is the recognition of the Arg-Gly-Asp (RGD) sequence, known to be present in the extracellular matrix components fibronectin, vitronectin, collagen, fibrinogen, and von Willebrand factor (Cheresh, 1991). The involvement of integrins in vascular proliferation, adhesion, and wound repair has been well documented. The adhesion receptor, integrin alphaVbeta3, appears to be selectively expressed on growing blood vessels and has been identified as a marker of angiogenic vascular tissue (Brooks, 1994). Due to its involvement in angiogenesis, integrin alphaVbeta3 is one of the most intensely studied of the integrin receptors.

Specificity

Reacts with the vitronectin receptor alphaVbeta3 complex, an RGD-directed adhesion receptor. LM609 has been demonstrated to block adhesion of a human melanoma cell line (M21) to vitronectin, fibrinogen and von Willebrand factor, as well as to a synthetic RGD containing peptide (Cheresh, 1987). In chick chorioallentoic membranes, LM609 was shown to block angiogenesis induced by bFGF and TNFalpha but had no effect on pre-existing vessels (Brooks, 1994). While LM609 does block cell attachment to RGD containing ligands, it does not interact directly with the RGD binding site. Instead, LM609 appears to be an allosteric inhibitor of integrin alphaVbeta3, which binds to a conformational epitope resulting from the post-translational association of the alphaV and beta3 subunits.

Immunogen

Purified adhesion receptor from the human melanoma cell line M21 (Cheresh, 1987).

Application

Anti-Integrin αVβ3 Antibody, clone LM609, FITC conjugated detects level of Integrin αVβ3 & has been published & validated for use in FC, IH.
Flow Cytometry: 1-3 μg LM609 per 0.5 x 10e6 cells in 100μL (Smith, 1996).

Immunofluorescence: 5-10 μg/mL for staining of bFGF-treated 6 mm cryosections of chick chorioallantoic membrane fixed with acetone (Brooks, 1994). LM609 is also effective on 4% paraformaldehyde fixed, frozen tissues and cells. However it is not effective for immunohistochemical staining of paraffin-embedded tissue sections.

Optimal working dilutions and protocols must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Liquid in 0.02M PB (pH 7.6), 0.25M NaCl, and 0.1% sodium azide with 15 mg/mL BSA as a stabilizer.
Protein A purified

Storage and Stability

Maintain at 2–8°C in undiluted aliquots for up to 6 months. Protect from Light. Do not store as a dilute solution.

Analysis Note

Control
POSITIVE CONTROL: Human myeloma cell lines M21 and MoalphaV (Chen, 1995).

NEGATIVE CONTROL: Mo cells (this cell line was derived from M21 but does not express integrin alphaV

Chen, 1995).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uri Abadi et al.
Journal of clinical medicine, 10(8) (2021-05-01)
Chronic lymphocytic leukemia (CLL) is the most common adult leukemia. The thyroid hormones, T3 and T4, bind the αvβ3 integrin and activate phosphorylates ERK (pERK). These tumor-promoting actions were reported in a number of malignancies, but not in CLL. Primary
Yi Liu et al.
International journal of cancer, 121(1), 184-192 (2007-03-03)
Unlike the intact fibronectin (FN) molecule, some proteolytic or recombinant fragments of FN possess inhibitory activities on tumor, providing potential strategies in tumor therapeutics. Using the hydrodynamics-based gene delivery technique, we demonstrated that the treatment by in vivo expression of
Michel Laguerre et al.
PloS one, 8(11), e78683-e78683 (2013-11-16)
Mutations in ITGA2B and ITGB3 cause Glanzmann thrombasthenia, an inherited bleeding disorder in which platelets fail to aggregate when stimulated. Whereas an absence of expression or qualitative defects of αIIbβ3 mainly affect platelets and megakaryocytes, αvβ3 has a widespread tissue
Gal Yosef et al.
The Journal of biological chemistry, 293(34), 13310-13326 (2018-07-11)
The pathophysiological functions of the signaling molecules matrix metalloproteinase-14 (MMP-14) and integrin αvβ3 in various types of cancer are believed to derive from their collaborative activity in promoting invasion, metastasis, and angiogenesis, as shown in vitro and in vivo The
Paul A Dayton et al.
Molecular imaging, 3(2), 125-134 (2004-08-07)
The goal of targeted ultrasound contrast agents is to significantly and selectively enhance the detection of a targeted vascular site. In this manuscript, three distinct contrast agents targeted to the alphavbeta3 integrin are examined. The alphavbeta3 integrin has been shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service