Skip to Content
Merck
All Photos(1)

Key Documents

CBL1300

Sigma-Aldrich

Anti-VCAM-1 Antibody, clone M/K

clone M/K - 2, Chemicon®, from rat

Synonym(s):

CD106

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

M/K - 2, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes Murine vascular adhesion molecule (VCAM-1), single chain glycoprotein of Mr 100- kDa (under reducing conditions). VCAM-1 is present on the +/+ 2.4 stromal cell clone, large cells in bone marrow, and a subset of thymic macrophages. The stromal cell antigen is also detectable on the EOMA and BMS2 endothelial cell lines. The antibody reacts with all known isoforms of murine VCAM-1.

Application

Anti-VCAM-1 Antibody, clone M/K is an antibody against VCAM-1 for use in FC, IH & IP.
Studies of cell-cell interaction

Cell adhesion assays

Inhibition of lymphopoieisis in Whitlock-Witte long-term marrow cultures

Immunoprecipitation of the VCAM-1 integrin complex

Detection of VCAM-1 cells by immunofluorescence and immunohistochemical staining: acetone or fresh frozen sections work best; although zinc formalin or PLP fixations are reported to work but they have not be verified by Chemicon.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: MAB2242

Physical form

Format: Purified

Analysis Note

Control
EOMA and BMS2 endothelial cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haowen Qiao et al.
Journal of medical virology, 96(5), e29671-e29671 (2024-05-15)
The coronavirus disease of 2019 (COVID-19) pandemic has led to more than 700 million confirmed cases and nearly 7 million deaths. Although severe acute respiratory syndrome coronavirus-2 (SARS-CoV-2) virus mainly infects the respiratory system, neurological complications are widely reported in
Michael J Borrett et al.
Cell reports, 32(6), 108022-108022 (2020-08-14)
The transitions from developing to adult quiescent and activated neural stem cells (NSCs) are not well understood. Here, we use single-cell transcriptional profiling and lineage tracing to characterize these transitions in the murine forebrain. We show that the two forebrain
Sophie F Ellermann et al.
Frontiers in pharmacology, 13, 992262-992262 (2022-12-20)
Major surgery induces systemic inflammation leading to pro-inflammatory activation of endothelial cells. Endothelial inflammation is one of the drivers of postoperative organ damage, including acute kidney injury Tumour Necrosis Factor alpha (TNF-α) is an important component of surgery-induced pro-inflammatory activation
Elham Khanicheh et al.
PloS one, 8(3), e58761-e58761 (2013-04-05)
Inflammatory changes on the endothelium are responsible for leukocyte recruitment to plaques in atherosclerosis. Noninvasive assessment of treatment-effects on endothelial inflammation may be of use for managing medical therapy and developing novel therapies. We hypothesized that molecular imaging of vascular
Cathy Grutzmacher et al.
American journal of physiology. Renal physiology, 304(1), F19-F30 (2012-10-19)
Diabetic nephropathy is the most common cause of end-stage renal disease and is a major risk factor for cardiovascular disease. In the United States, microvascular complications during diabetic nephropathy contribute to high morbidity and mortality rates. However, the cell-autonomous impact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service