Skip to Content
Merck
All Photos(1)

Key Documents

AR00206

Millipore

VIRAPREP Kit

new

ISO 10705-2, suitable for filter test (Somatic coliphages and F-specific RNA bacteriophages)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.84

Agency

ISO 10705-2

shelf life

12 mo.

packaging

pack of 100 tests

suitability

suitable for filter test (Somatic coliphages and F-specific RNA bacteriophages)

application(s)

water monitoring

storage temp.

2-8°C

General description

VIRAPREP Kit is designed to screen 100 test samples through filtration method with sample volume ranging between 10-100ml. The kit targets screening of somatic coliphages (PhiX174 model) and F-specific RNA bacteriophages (MS2 model).
The detection limit of 1 PFU/100ml is in accordance with EU requirements.

Application

VIRAPREP Kit is used to concentrate and elute somatic coliphages and F-specific RNA bacteriophages from drinking water and treated wastewater samples.

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service