Skip to Content
Merck
All Photos(1)

Key Documents

8.21036

Sigma-Aldrich

Piperine

for synthesis

Synonym(s):

Piperine, 1-Piperoylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-348-0
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

potency

514 mg/kg LD50, oral (Rat)

mp

127-130 °C

solubility

0.04 g/L

storage temp.

2-30°C

InChI

1S/C17H19NO3/c19-17(18-10-4-1-5-11-18)7-3-2-6-14-8-9-15-16(12-14)21-13-20-15/h2-3,6-9,12H,1,4-5,10-11,13H2/b6-2+,7-3+

InChI key

MXXWOMGUGJBKIW-YPCIICBESA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 128 °C
Melting range (upper value): ≤ 131 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service