Skip to Content
Merck
All Photos(1)

Key Documents

8.20099

Sigma-Aldrich

Anilinium chloride

for synthesis

Synonym(s):

Anilinium chloride, Aniline hydrochloride, Aniline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N · xHCl
CAS Number:
Molecular Weight:
93.13 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-519-8
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

solid

potency

840 mg/kg LD50, oral (Rat)

bp

245 °C/1013 hPa

mp

199-202 °C

transition temp

flash point 193 °C

solubility

1070 g/L

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].[N+H3]c1ccccc1

InChI

1S/C6H7N.ClH/c7-6-4-2-1-3-5-6;/h1-5H,7H2;1H

InChI key

MMCPOSDMTGQNKG-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Melting range (lower value): ≥ 196 °C
Melting range (upper value): ≤ 201 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

381.2 °F - closed cup

Flash Point(C)

194 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service