Skip to Content
Merck
All Photos(1)

Key Documents

616452

Sigma-Aldrich

RepSox

≥95% (HPLC), solid, TGF-β RI kinase inhibitor, Calbiochem

Synonym(s):

TGF-β RI Kinase Inhibitor II, ALK5 Inhibitor II, Transforming Growth Factor-β Type I Receptor Kinase Inhibitor II, 2-(3-(6-Methylpyridin-2-yl)-1H-pyrazol-4-yl)-1,5-naphthyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N5
CAS Number:
Molecular Weight:
287.32
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

TGF-β RI Kinase Inhibitor II, TGF-β RI Kinase Inhibitor II, CAS 446859-33-2, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of TGF-β R1 kinase (IC₅₀ = 23 nM and 4 nM for ALK5 binding & auto-phosphorylation).

Quality Level

Assay

≥95% (HPLC)

form

solid

potency

23 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1ncc(c1c4nc(ccc4)C)c2nc3c(nccc3)cc2

InChI

1S/C17H13N5/c1-11-4-2-5-16(20-11)17-12(10-19-22-17)13-7-8-14-15(21-13)6-3-9-18-14/h2-10H,1H3,(H,19,22)

InChI key

LBPKYPYHDKKRFS-UHFFFAOYSA-N

General description

A cell-permeable naphthyridinyl pyrazolo compound that acts as a potent, selective, reversible, and ATP-competitive inhibitor of TGF-β type I receptor (ALK5; IC50 = 23 nM, 4 nM and 18 nM for ALK5 binding, ALK5 auto-phosphorylation and TGF-β cellular assay in HepG2 cells, respectively). Minimally affects a panel of 9 closely related kinases including p38 MAPK at IC50 >16 µM. Also available as a 50 mM solution in DMSO (Cat. No. 508158).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ALKS binding
Product competes with ATP.
Secondary Target
ALK 5 auto phosphorylation (IC₅₀ = 4 nM)

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Gellibert, F., et al. 2004. J. Med. Chem.47, 4494.
Ichida, J. K., et al. 2009. Cell Stem Cell 5, 491.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ayumi Kanemaru et al.
Cancer cell international, 22(1), 358-358 (2022-11-16)
Tumor suppressor CYLD dysfunction by loss of its expression, triggers malignant transformation, especially drug resistance and tumor invasion/metastasis. Although loss of CYLD expression is significantly associated with poor prognosis in a large variety of tumors, no clinically-effective treatment for CYLD-negative
Satoru Shinriki et al.
The Journal of pathology, 244(3), 367-379 (2017-12-14)
Oral squamous cell carcinoma (OSCC) has a very poor prognosis because of its highly invasive nature, and the 5-year survival rate has not changed appreciably for the past 30 years. Although cylindromatosis (CYLD), a deubiquitinating enzyme, is thought to be a
Mikolaj M Kozlowski et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(13), 2618-2632 (2020-02-23)
Sensory hair cell losses underlie the vast majority of permanent hearing and balance deficits in humans, but many nonmammalian vertebrates can fully recover from hearing impairments and balance dysfunctions because supporting cells (SCs) in their ears retain lifelong regenerative capacities
Ana Guerrero et al.
Nature aging, 2, 851-866 (2022-11-29)
Cellular senescence is a stable type of cell cycle arrest triggered by different stresses. As such, senescence drives age-related diseases and curbs cellular replicative potential. Here, we show that 3-deazaadenosine (3DA), an S-adenosyl homocysteinase (AHCY) inhibitor, alleviates replicative and oncogene-induced
Sayaka Imatsuji et al.
Oncology research, 32(1), 139-150 (2024-01-08)
Growing evidence suggests an association between epithelial-mesenchymal transition (EMT), a hallmark of tumor malignancy, and chemoresistance to a number of anti-cancer drugs. However, the mechanism of EMT induction in the process of acquiring anti-cancer drug resistance remains unclear. To address

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service