Skip to Content
Merck
All Photos(1)

Key Documents

557403

Sigma-Aldrich

RNA Polymerase III Inhibitor

RNA Polymerase III Inhibitor, CAS 577784-91-9, is a cell-permeable inhibitor of RNA Polymerase III (IC₅₀ = 27 and 32 µM for human and S. cerevisiae RNA Pol III, respectively).

Synonym(s):

RNA Polymerase III Inhibitor, ML-60218, N-[1-(3-(5-Chloro-3-methylbenzo[b]thiophen-2-yl-1-methyl-1H-pyrazol-5-yl)]-2-chlorobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15Cl2N3O2S2
CAS Number:
Molecular Weight:
452.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 15 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Nc2[n](nc(c2)c3[s]c4c(c3C)cc(cc4)Cl)C)c1c(cccc1)Cl

InChI

1S/C19H15Cl2N3O2S2/c1-11-13-9-12(20)7-8-16(13)27-19(11)15-10-18(24(2)22-15)23-28(25,26)17-6-4-3-5-14(17)21/h3-10,23H,1-2H3

InChI key

BVBDTTLISMIOJY-UHFFFAOYSA-N

General description

A cell-permeable indazolo-sulfonamide compound that displays broad-spectrum inhibitory activity against RNA Polymerase III (IC50 = 27 and 32 µM for human and S. cerevisiae RNA Pol III, respectively). Inhibits cell growth in yeast by preventing RNA Pol III-mediated tRNA transcription. Also available as a 25 mM solution in DMSO (Cat. No. 557404).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
activity against RNA Polymerase III
Product does not compete with ATP.
Reversible: no
Target IC50: 27 and 32 µM for Human and S. cerevisiae RNA Pol III

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Preparation Note

Slight warming may be necessary to achieve complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wu, L., et al. 2003. Euk. Cell2, 256.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica M Silva et al.
RNA biology, 8(3), 496-505 (2011-05-03)
More than 98% of the human genome is comprised of non-protein coding sequences. Interestingly, a considerable fraction of these sequences is transcribed into non-protein coding RNA transcripts. These transcripts range in size from very small RNAs such as the miRNAs
Kevin Van Bortle et al.
Nature communications, 13(1), 3007-3007 (2022-06-01)
RNA polymerase III (Pol III) includes two alternate isoforms, defined by mutually exclusive incorporation of subunit POLR3G (RPC7α) or POLR3GL (RPC7β), in mammals. The contributions of POLR3G and POLR3GL to transcription potential has remained poorly defined. Here, we discover that
Maiko Kitaoka et al.
Current biology : CB, 32(18), 3939-3951 (2022-08-17)
Although central to evolution, the causes of hybrid inviability that drive reproductive isolation are poorly understood. Embryonic lethality occurs when the eggs of the frog X. tropicalis are fertilized with either X. laevis or X. borealis sperm. We observed that distinct subsets of
Magdalena Podlacha et al.
Nature communications, 15(1), 2274-2274 (2024-03-14)
One of the hopes for overcoming the antibiotic resistance crisis is the use of bacteriophages to combat bacterial infections, the so-called phage therapy. This therapeutic approach is generally believed to be safe for humans and animals as phages should infect
Jeffrey Y Zhou et al.
Proceedings of the National Academy of Sciences of the United States of America, 120(5), e2213777120-e2213777120 (2023-01-25)
The accrual of cytosolic DNA leads to transcription of type I IFNs, proteolytic maturation of the IL-1 family of cytokines, and pyroptotic cell death. Caspase-1 cleaves pro-IL1β to generate mature bioactive cytokine and gasdermin D which facilitates IL-1 release and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service