Skip to Content
Merck
All Photos(1)

Key Documents

539514

Sigma-Aldrich

Protein Phosphatase, Lambda, Recombinant, E. coli

Synonym(s):

Protein Phosphatase, Lambda

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥400,000 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

Protease, none detected

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant λ PPase expressed in E. coli. A 221-amino acid, Mn2+-dependent, protein phosphatase. Hydrolyzes phosphate groups on serine, threonine, tyrosine, or histidine residues. Inhibited by orthovanadate. Note: 1KU = 1,000 units.
Research Area: Cell Signaling
The protein phosphatase encoded by bacteriophage λ (λPP) is part of a family of Ser/Thr phosphatases (Ser/Thr PPases), which includes eukaryotic protein phosphatases 1 (PP1), 2A (PP2A), and 2B (calcineurin).

Application

Protein Phosphatase, Lambda, Recombinant, E. coli is suitable for the detection of phospho-KRAS.

Biochem/physiol Actions

Protein phosphatases play a crucial role in embryonic development, metabolic homeostasis, stress responses, cell cycle transitions, and numerous other vital biological processes.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 nmol of pNPP per min at 30°C, pH 7.8. 20-200 Units will typically remove >95% of phosphates from serine/threonine residues in 0.1 nmol of protein in 30 min. 100-1000 Units will typically remove >95% of phosphates from tyrosine in 0.1 nmol of protein in 30 min.

Physical form

In 100 mM NaCl, 50 mM HEPES, 2 mM DTT, 0.1 mM MnCl₂, 0.1 mM EGTA, 50% glycerol, 0.01% Brij® 35 Detergent, pH 7.5.

Preparation Note

Further dilute in 50 mM Tris-HCl, pH 7.8, 5 mM DTT, 2 mM MnCl₂, and 100 µg/ml BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Zhuo, S., et al. 1993. J. Biol. Chem.268, 17754.
Gordon, J.A. 1991. Methods Enzymol.201, 477.
Cohen, P.T.W. and Cohen, P. 1989. Biochem. J.260, 931.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorylation at Ser-181 of oncogenic KRAS is required for tumor growth
Barcel'o C, et al.
Cancer research, 74(4), 1190-1199 (2014)
Structure of the bacteriophage lambda Ser/Thr protein phosphatase with sulfate ion bound in two coordination modes
Voegtli WC, et al.
Biochemistry, 39(50), 15365-15374 (2000)
Optical control of protein phosphatase function
Courtney TM and Deiters A
Nature Communications, 10(1), 4384-4384 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service