Skip to Content
Merck
All Photos(1)

Documents

5.31067

Sigma-Aldrich

HSP70 Inhibitor, PES-Cl

Synonym(s):

HSP70 Inhibitor, PES-Cl, Heat Shock Protein 70 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6ClNO2S
CAS Number:
Molecular Weight:
215.66
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

2-5 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable chlorophenylethynylsulfonamide compound that binds directly to the substrate-binding domain of heat shock protein 70 (HSP70) and disrupts its interaction with client proteins. Impairs the activity of the anaphase promoting complex/cyclosome (APC/C) in cell-free extracts. Arrests cell cycle at G2/M phase and preferentially induces genomic instability in cancer cells (IC50 = 2 to 5 µM in melanoma cell lines). Exhibits much reduced toxicity in non-transformed cells (IC50 >100 µM in primary melanocytes). Induces apoptosis and inhibits autophagy as evidenced by increased p62SQSTM1 and LC3 accumulation. Also, shown to extend life in murine models of pre-B cell lymphoma.
A cell-permeable chlorophenylethynylsulfonamide compound that binds directly to the substrate-binding domain of heat shock protein 70 (HSP70) and disrupts its interaction with client proteins. Impairs the activity of the anaphase promoting complex/cyclosome (APC/C) in cell-free extracts. Arrests cell cycle at G2/M phase and preferentially induces genomic instability in cancer cells (IC50 = 2 to 5 µM in melanoma cell lines). Exhibits much reduced toxicity in non-transformed cells (IC50 >100 µM in primary melanocytes). Induces apoptosis and inhibits autophagy as evidenced by increased p62SQSTM1 and LC3 accumulation. Also, shown to extend life in murine models of pre-B cell lymphoma.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HSP70
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Balaburski, G., et al. 2013. Mol. Cancer Res.11, 1.
Budina-Kolomets, A. et al. 2013. Cancer Biol. Ther.15, 194.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anna Budina-Kolomets et al.
Cancer biology & therapy, 15(2), 194-199 (2013-10-09)
The chaperone HSP70 promotes the survival of cells exposed to many different types of stresses, and is also potently anti-apoptotic. The major stress-induced form of this protein, HSP70-1, is overexpressed in a number of human cancers, yet is negligibly expressed
Gregor M Balaburski et al.
Molecular cancer research : MCR, 11(3), 219-229 (2013-01-11)
The stress-induced HSP70 is an ATP-dependent molecular chaperone that plays a key role in refolding misfolded proteins and promoting cell survival following stress. HSP70 is marginally expressed in nontransformed cells, but is greatly overexpressed in tumor cells. Silencing HSP70 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service