Skip to Content
Merck
All Photos(1)

Key Documents

5.30583

Sigma-Aldrich

α-Synuclein Blocker, ELN484228

Synonym(s):

α-Synuclein Blocker, ELN484228, N-(4-Fluorophenyl)benzenesulfonamide, NSC164389, ELN 484228

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10FNO2S
CAS Number:
Molecular Weight:
251.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

InChI

1S/C12H10FNO2S/c13-10-6-8-11(9-7-10)14-17(15,16)12-4-2-1-3-5-12/h1-9,14H

InChI key

AZORQGXJAXEIGC-UHFFFAOYSA-N

General description

A cell-permeable phenylsulfonamide compound that reversibly binds to pocket I of the monomeric α-synuclein (α-syn) and reverses α-syn-induced impairment of phagocytosis in H4 neuroglioma cells over-expressing wild-type α-syn and microglia isolated from α-syn BAC transgenic animals over-expressing the E46K mutant form of human α-syn (~1-10 µM). Reduces α-syn-mediated impairment of vesicular dynamics and diminishes α-syn translocation to the synaptic region in rat primary neuronal cultures by 50% (at ~ 1 µM) and in cortical cultures in transgenic mice over-expressing human α-syn. Also shown to protect dopaminergic neurons against the toxic effects of over-expressed α-syn A53T and reduces neurite retraction in TH- and MAP2-positive neurons.
A cell-permeable phenylsulfonamide compound that reversibly binds to pocket I of the monomeric α-synuclein (α-syn) and reverses α-syn-induced impairment of phagocytosis in H4 neuroglioma cells over-expressing wild-type α-syn and microglia isolated from α-syn BAC transgenic animals over-expressing the E46K mutant form of human α-syn (~1-10 µM). Reduces α-syn-mediated impairment of vesicular dynamics and diminishes α-syn translocation to the synaptic region in rat primary neuronal cultures by 50% (at ~ 1 µM) and in cortical cultures in transgenic mice over-expressing human α-syn. Also shown to protect dopaminergic neurons against the toxic effects of over-expressed α-syn A53T and reduces neurite retraction in TH- and MAP2-positive neurons.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
monomeric α-synuclein
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Toth, G., et al. 2014. PLoS ONE9, e87133.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service