Skip to Content
Merck
All Photos(1)

Key Documents

239807

Sigma-Aldrich

Cyclopamine-KAAD

≥70% (sum of two isomers, HPLC), liquid, Hedgehog signaling inhibitor, Calbiochem®

Synonym(s):

3- Keto-N-( aminoethyl- aminocaproyl- dihydrocinnamoyl)cyclopamine, 3-Keto-N-(aminoethyl-aminocaproyl-dihydrocinnamoyl)cyclopamine, InSolution Cyclopamine-KAAD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.77

product name

Cyclopamine-KAAD, InSolution, ≥70%, inhibits Hedgehog (Hh) signaling

Quality Level

Assay

≥70% (sum of two isomers, HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

dry ice

storage temp.

−70°C

General description

A cell-permeable potent analog of Cyclopamine (>Cat. No. 239803) that specifically inhibits Hedgehog (Hh) signaling with similar or lower toxicity (IC50 = 20 nM in Shh-LIGHT2 assay; 50 nM in p2Ptch-/-cells; 500 nM in SmoA1-LIGHT cells). Binds to SmoA1 and promotes its exit from the endoplasmic reticulum. Suppresses both ShhNp-induced pathway activity and SmoA1-induced reporter activity. Reported to cause regression of murine tumor allografts in vivo and induce rapid cell death in human medulloblastoma. Shown to sensitize human glioma cells to TRAIL-induced apoptosis. The solid form of this compound (Cat. No. 239804) is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
SmoA1

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 1 mM (50 µg/72 µL) solution of Cyclopamine-KAAD (Cat. No. 239804) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C.

Other Notes

Siegelin, M.D. et al. 2009. Neurobiol. Dis.34, 259.
Watkins, D.N., et al. 2003. Nature422, 313.
Berman, D.M., et al. 2002. Science297, 1559.
Chen, J.K., et al. 2002. Proc. Natl. Acad. Sci. USA99, 14071.
Chen, J.K., et al. 2002. Genes Dev.16, 2743.
Frank-Kamenetsky, M., et al. 2002. J. Biol.1, 10.
Taipale, J., et al. 2000. Nature406, 1005.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Betty R Lawton et al.
Endocrinology, 161(10) (2020-08-19)
Differentiation of pluripotent stem cells into functional parathyroid-like cells would accelerate development of important therapeutic options for subjects with parathyroid-related disorders, from the design and screening of novel pharmaceutical agents to the development of durable cellular therapies. We have established

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service