Skip to Content
Merck
All Photos(2)

Documents

1.51453

Supelco

Purospher®STAR RP-8 endcapped (5 µm) HPLC Columns

L × I.D. 150 mm x 4.6 mm Hibar® RT HPLC Column

Synonym(s):

Purospher® STAR RP-8 endcapped (5 µm) Hibar® RT 150-4.6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

Purospher® STAR RP-8 endcapped (5 µm), L × I.D. 150 mm x 4.6 mm Hibar® RT HPLC column

Quality Level

feature

endcapped

parameter

400 bar pressure
65 °C max. temp.

technique(s)

HPLC: suitable
mass spectrometry (MS): suitable (High)

matrix

fully porous particle

particle size

5 μm

pore size

120 Å pore size

operating pH range

1.5-10.5

separation technique

supercritical fluid (SFC)

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

General description

HPLC column
Purospher® STAR RP-8 endcapped allows best results and highest flexibility due to its widest pH range up to pH 10.5, best performance and separation highest efficiency. Purospher STAR RP-8 endcapped is suited very well for the separation of strong hydrophobic compounds and peptides.

Analysis Note

Pressure: ≤ 80 bar
Theoretical Plates (N/m) (Toluene): ≥ 85000
Symmetry (Toluene): 0.8 - 1.4
Capacity factor (Toluene): ≥ 1.0

Legal Information

Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service