Skip to Content
Merck
All Photos(1)

Key Documents

1.06627

Supelco

Sodium thiocyanate

EMPLURA®

Synonym(s):

Sodium thiocyanate, Sodium sulfocyanide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CNNaS
CAS Number:
Molecular Weight:
81.07
MDL number:
UNSPSC Code:
12352302
EC Index Number:
208-754-4
NACRES:
NA.21
Assay:
≥98.5% dry basis (silver nitrate titration)
form:
solid
solubility:
>1000 g/L
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥98.5% dry basis (silver nitrate titration)

form

solid

potency

764 mg/kg LD50, oral (Rat)

loss

≤5.0% loss on drying, 105°C

pH

6-8 (20 °C, 100 g/L in H2O)

mp

308 °C

solubility

>1000 g/L

density

1.74 g/cm3 at 20 °C

bulk density

800‑1000 kg/m3

anion traces

chloride (Cl-): ≤0.1%
sulfate (SO42-): ≤0.025%

cation traces

As: ≤0.0005%
heavy metals (as Pb): ≤0.002%

storage temp.

2-30°C

SMILES string

[Na+].[S-]C#N

InChI

1S/CHNS.Na/c2-1-3;/h3H;/q;+1/p-1

InChI key

VGTPCRGMBIAPIM-UHFFFAOYSA-M

Related Categories

Application


  • Copper-mediated [3 + 2] oxidative cyclization of oxime acetate: Demonstrates the use of sodium thiocyanate in copper-mediated synthesis, crucial for the formal synthesis of fentiazac, highlighting its role in pharmaceutical synthesis techniques (Jadhao et al., 2024).

  • Fabrication and Characterization of Flexible Solid Polymers Electrolytes for Supercapacitor Application: Details the application of sodium thiocyanate in the development of advanced polymer electrolytes for supercapacitors, emphasizing its impact on the energy storage sector (Badi et al., 2022).

  • Lactose oxidase: An enzymatic approach to inhibit Listeria monocytogenes in milk: Utilizes sodium thiocyanate as part of a novel enzymatic strategy to enhance food safety, providing an alternative to traditional preservatives in dairy products (Flynn et al., 2021).

  • 1,3-Dibromo-1,1-difluoro-2-propanone as a Useful Synthon for a Chemoselective Preparation of 4-Bromodifluoromethyl Thiazoles: Incorporates sodium thiocyanate in the synthesis of bromodifluoromethyl thiazoles, demonstrating its utility in the creation of novel compounds for chemical research and industrial applications (Colella et al., 2018).

Analysis Note

Assay (argentometric; calculated on dried substance): ≥ 98.5 %
Identity: passes test
Chloride (Cl): ≤ 0.1 %
Sulfate (SO₄): ≤ 0.025 %
Heavy metals (as Pb): ≤ 0.002 %
As (Arsenic): ≤ 0.0005 %
Loss on Drying (105°C): ≤ 5.0 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service