Skip to Content
Merck
All Photos(2)

Documents

1.00658

Supelco

Acetone

for gas chromatography MS SupraSolv®

Synonym(s):

Propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-662-2
NACRES:
NA.03

vapor density

2 (vs air)

Quality Level

vapor pressure

184 mmHg ( 20 °C)

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

465 °C (DIN 51794)

potency

5800 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

expl. lim.

13.2 %

technique(s)

GC/MS: suitable

impurities

≤0.05% Water

evapn. residue

≤3.0 mg/L

color

APHA: ≤10

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L in H2O)

capacity

1000  g/kg water absorbing capacity

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

transition temp

flash point <-20 °C

density

0.791 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Pesticide analysis, SPME Fiber, supelco spme fiber, direct immersion spme, pesticide analysis methods, pesticide analysis by gc-ms, DI-SPME, overcoated SPME, overcoated fibers, soy milk analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service