Skip to Content
Merck
All Photos(1)

Key Documents

1.00629

Supelco

Rubeanic acid

GR for analysis

Synonym(s):

Rubeanic acid, Dithiooxamide, Rubean-hydrogen, Ethanedithioamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4N2S2
CAS Number:
Molecular Weight:
120.20
MDL number:
UNSPSC Code:
12352111
EC Index Number:
201-203-9

Quality Level

form

solid

potency

500 mg/kg LD50, oral (Rat)

mp

200 °C (decomposition)

solubility

0.37 g/L

bulk density

650 kg/m3

storage temp.

15-25°C

InChI

1S/C2H4N2S2/c3-1(5)2(4)6/h(H2,3,5)(H2,4,6)

InChI key

OAEGRYMCJYIXQT-UHFFFAOYSA-N

Analysis Note

Assay (alkalimetric): ≥ 98.0 %
Identity (IR-spectrum): conforms
Suitability for copper determination: conforms

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service