Skip to Content
Merck
All Photos(2)

Key Documents

07-081

Sigma-Aldrich

Anti-phospho (Ser10)-acetyl (Lys14)-Histone H3 Antibody

Upstate®, from rabbit

Synonym(s):

H3K14me3S10P, Histone H3 (acetyl K14, phospho S10), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

eukaryotes, mouse, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer10), acetylation (Lys14)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3.1 (UniProt: Q16695; also known as H3K14AcS10P, Histone H3 (acetyl K14, phospho S10), H3 histone family member T, Histone 3, H3, Histone cluster 3, H3) is encoded by the HIST3H3 (also known as HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ, HIST3H3, H3FT, H3T, MGC126886) gene (Gene ID: 8350, 8351, 8352, 8353, 8354, 8355, 8356, 8357, 8358, 8968) in human. Histone H3 has two main variants, H3.1 and H3.3, which show different genomic localization patterns in animals. The H3.1 and H3.3 complexes also possess distinct histone chaperones, CAF-1 and HIRA, which play important role in mediating DNA-synthesis-dependent and -independent nucleosome assembly. It has been reported that Histone H3.1 serves as the canonical histone, which is incorporated during DNA replication, whereas H3.3 acts as the replacement histone that can be incorporated outside of S-phase during chromatin-disrupting processes like transcription. Histone H 3.1 is a core component of nucleosome that is present only in mammals and is usually enriched in acetylation of Lysine 15 and demethylation of lysine 10 (HeK9Me2). It is expressed during S phase, then expression decreases significantly as cell division slows down during the process of differentiation. Histone H 3.1 expression is shown to be replication dependent. It s presence at the site of UV-induced DNA damage has also been reported. It has also been shown that H3.1/H4 tetramers do not split and remain intact during replication dependent deposition of H3.1 variant. Phosphorylation at serine 10 by aurora kinase B is reported to mediate the dissociation of HP1 proteins (CBX1, CBX3 and CBX5) from heterochromatin and this phosphorylation is considered to be an essential regulatory mechanism for neoplastic cell transformation. Acetylation of lysine 14 of histone H3 (H3K14ac) is shown to be critical for DNA damage checkpoint activation by directly regulating the compaction of chromatin and by recruiting chromatin remodeling protein complex RSC (Remodels the Structure of Chromatin). Nucleosomes with H3K14ac display higher affinity for purified RSC. (Ref.: Stroud, H., et al (2012). Proc. Natl. Acad. Sci. USA 109(14); 5370-75; Dunn, MR., and Smerdon MJ (2014). J. Biol. Chem. 289(12);8353-63)..

Specificity

Broad species cross-reactivity expected.
Recognizes Histone H3 phosphorylated at serine 10 and acetylated at lysine 14, Mr 17 kDa. An unidentified protein is also seen in some cells lines, Mr 40 kDa.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 7-20 of human histone H3 (ARK[pS]TGGAcKAPRKQL-C).

Application

Anti-phospho (Ser10)-acetyl (Lys14)-Histone H3, Cat. No. 07-081, is a highly specific rabbit polyclonal antibody that targets Histone H3 phosphorylated on Serine 10 and acetylated on Lysine 14 and has been tested for use in Immunocytochemistry, Peptide Inhibition Assay, ChIP-seq and Chromatin Immuno
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western Blot Analysis:
1:2000-1:10,000 dilutions of a previous lot detected phosphorylated and acetylated histone H3 in acid extracted proteins from serum starved mouse 10T1/2 cells treated with 50ng/ml EGF for 12.5 minutes. Western blotting of this lot to acid extracted proteins described above was preferentially competed by peptides in the following order: phospho-acetyl-H3 (7-20) > straight chain H3 (7-20) > phospho-H3 (7-20) or acetyl-H3 (7-20).
Peptide Inhibition Analysis: A 1:500 dilution from a representative lot was used with HeLa AE for peptide block analysis.
Chromatin Immunoprecipitation:
A previous lot of this antibody was reported by an independent laboratory to immunoprecipitate chromatin. (Cheung, P., 2000.)

Immunocytochemistry:
A previous lot of this antibody detected phosphorylated and acetylated histone H3 in EGF-stimulated cells; as reported by an independent laboratory. (Cheung, P., 2000.)

Quality

Routinely evaluated by Western Blot on HeLa acid extract.

Western Blot Analysis:
1:500 dilution of this lot detected phospho acetyl Histone H3 on 10 μg of HeLa acid extract.

Target description

17 kDa

Physical form

Immunoaffinity and reverse-affinity chromatography.
Purified rabbit polyclonal IgG in buffer containing 0.2 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide, before the addition of glycerol to 30%.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Acid-extracted proteins from serum-starved 10T1/2 cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BIR-1, a Caenorhabditis elegans homologue of Survivin, regulates transcription and development.
Kostrouchova, M; Kostrouch, Z; Saudek, V; Piatigorsky, J; Rall, JE
Proceedings of the National Academy of Sciences of the USA null
Lovastatin-induced cholesterol depletion affects both apical sorting and endocytosis of aquaporin-2 in renal cells.
Procino G, Barbieri C, Carmosino M, Rizzo F, Valenti G, Svelto M
American Journal of Physiology: Renal Physiology null
Targeted expression of cyclin D2 results in cardiomyocyte DNA synthesis and infarct regression in transgenic mice.
Pasumarthi, KB; Nakajima, H; Nakajima, HO; Soonpaa, MH; Field, LJ
Circulation Research null
Beatriz Dominguez Gonzalez et al.
Human molecular genetics, 27(12), 2154-2170 (2018-06-06)
Heterozygosity for the TOR1A-Δgag mutation causes semi-penetrant childhood-onset dystonia (OMIM #128100). More recently, homozygous TOR1A mutations were shown to cause severe neurological dysfunction in infants. However, there is little known about the recessive cases, including whether existing reports define the
p38-Dependent marking of inflammatory genes for increased NF-kappa B recruitment
Saccani, S., et al
Nature Immunology, 3, 69-75 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service