Skip to Content
Merck
All Photos(3)

Key Documents

06-734

Sigma-Aldrich

Anti-β-Catenin Antibody

Upstate®, from rabbit

Synonym(s):

catenin (cadherin-associated protein), beta 1 (88kD), catenin (cadherin-associated protein), beta 1 (88kDa), catenin (cadherin-associated protein), beta 1, 88kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, rabbit, bovine, mouse

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CTNNB1(1499)
mouse ... Ctnnb1(12387)
rat ... Ctnnb1(84353)

General description

β-catenin plays a central role in cell adhesion, bridging between cadherins and actin cytoskeleton. These activities are regulated through its differential interaction with several molecular partners, including cell-cell adherens junction (AJ) molecules, components of its degradation complex, and transcription factors.
β-Catenin is composed of three domains: a central armadillo repeat domain and two N- and C-terminal tails. β-catenin can be phosphorylated by serine-threonine kinase, such as GSK-3. The phosphorylated β-catenin affects the protein association of β-catenin to E-cadherin and α-cateinin that occurs in the cell. This process can be reversed by de-phosphorylation of β-catenin.

Specificity

Recognizes β-Catenin, Mr 92 kDa.

Immunogen

KLH-conjugated, synthetic peptide (CGG-SYLDSGIHSGATTTAPSLSGK) corresponding to the consensus GSK3 phosphorylation site of human β -Catenin (amino acids 29-49).

Application

Immunoprecipitation:
5 μg of a previous lot immunoprecipitated β-Catenin from 500 μg of A431 membrane fraction lysate.

Immunocytochemistry:
1-5 μg/mL of a previous lot showed positive immunostaining for β-catenin at cell-cell junctions of A431 cells fixed with 4% paraformaldehyde and permeabilized with 0.25% Triton X-100.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected beta-Catenin in A431 and HUVEC cell lines.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-β-Catenin Antibody (Rabbit Polyclonal Antibody) validated in ICC, IP, WB to detect β-Catenin also known as catenin (cadherin-associated protein) beta 1 (88kD).

Quality

Routinely evaluated by western blot on A431 total cell lysate.

Western Blot Analysis:
0.1-1 μg/mL of this lot detected β-catenin in an A431 total cell lysate.

Target description

92 kDa

Physical form

Format: Purified
Protein A chromatography
Purified rabbit polyclonal IgG in buffer containing 0.1M Tris Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Handling Recommendations: Prior to removing the cap, centrifuge the vial and gently mix the solution.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

beta-Catenin/TCF/LEF regulate expression of the short form human Cripto-1.
Hamada, S; Watanabe, K; Hirota, M; Bianco, C; Strizzi, L; Mancino, M; Gonzales, M; Salomon, DS
Biochemical and biophysical research communications null
Beta-catenin-induced melanoma growth requires the downstream target Microphthalmia-associated transcription factor.
Widlund, HR; Horstmann, MA; Price, ER; Cui, J; Lessnick, SL; Wu, M; He, X; Fisher, DE
The Journal of cell biology null
Differential trafficking of transforming growth factor-beta receptors and ligand in polarized epithelial cells.
Murphy, SJ; Dore, JJ; Edens, M; Coffey, RJ; Barnard, JA; Mitchell, H; Wilkes, M; Leof, EB
Molecular Biology of the Cell null
Adherens junction proteins in the hamster uterus: their contributions to the success of implantation.
Luan, L; Ding, T; Stinnett, A; Reese, J; Paria, BC
Biology of reproduction null
Cooperative control via lymphoid enhancer factor 1/T cell factor 3 and estrogen receptor-alpha for uterine gene regulation by estrogen.
Ray, S; Xu, F; Wang, H; Das, SK
Molecular Endocrinology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service