Skip to Content
Merck
All Photos(1)

Documents

05-932

Sigma-Aldrich

Anti-Calmodulin Binding Protein Epitope Tag Antibody, clone C16T, rabbit monoclonal

clone C16T, Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C16T, monoclonal

species reactivity

vertebrates

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CALM1(801)

Specificity

Recombinant proteins containing the calmodulin binding protein epitope tag.

Immunogen

KLH-conjugated, synthetic peptide corresponding to the Calmodulin Binding Protein (CBP) epitope tag found in several E. coli expression vectors

Application

Anti-Calmodulin Binding Protein Epitope Tag Antibody, clone C16T is an antibody against Calmodulin Binding Protein Epitope Tag for use in WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

Routinely evaluated by immunoblot on E. coli transformed with a plasmid encoding a calmodulin binding protein epitope tagged protein

Target description

The molecular weight is related to the tagged protein.

Physical form

Format: Purified
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Samples containing the calmodulin binding protein epitope tag.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modular binding domains in signal transduction proteins.
Cohen, G B, et al.
Cell, 80, 237-248 (1995)
Affinity purification of histidine-tagged proteins.
Schmitt, J, et al.
Molecular Biology Reports, 18, 223-230 (1993)
Strategies for the identification of interacting proteins.
Guarente, L
Proceedings of the National Academy of Sciences of the USA, 90, 1639-1641 (1993)
D W Carr et al.
The Journal of biological chemistry, 266(22), 14188-14192 (1991-08-05)
The type II cAMP-dependent protein kinase is localized to specific subcellular environments through the binding of the regulatory subunit (RII) dimer to RII-anchoring proteins. Computer-aided analysis of secondary structure, performed on four RII-anchoring protein sequences (the microtubule-associated protein 2, P150
A single step purification for recombinant proteins. Characterization of a microtubule associated protein (MAP 2) fragment which associates with the type II cAMP-dependent protein kinase.
Stofko-Hahn, R E, et al.
Febs Letters, 302, 274-278 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service