Skip to Content
Merck
All Photos(2)

Key Documents

05-740-AF488

Sigma-Aldrich

Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12, Alexa Fluor 488

clone 10H11.E12, from mouse, ALEXA FLUOR 488

Synonym(s):

Serine-protein kinase ATM, Ser1981-phosphorylated, Ataxia telangiectasia mutated, Ser1981-phosphorylated, A-T mutated, Ser1981-phosphorylated, ATM, Ser1981-phosphorylated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

10H11.E12, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pSer1981)

Gene Information

human ... ATM(472)

General description

Serine-protein kinase ATM (EC 2.7.11.1; UniProt Q13315; also known as A-T mutated, Ataxia telangiectasia mutated) is encoded by the ATM (also known as AT) gene (Gene ID 472) in human. Ataxia telangiectasia mutated kinase (ATM) and ataxia telangiectasia and Rad3-related kinase (ATR) are related kinases that regulate cell cycle checkpoints and DNA repair. Mutation in the ATM gene results in the autosomal recessive disease ataxia telangiectasia (AT). Known ATM substrates include p53, p95/NBS1, MDM2, Chk2, BRCA1, CtIP, 4E-BP1, and Chk1. The S/TQ sequence constitutes the essential substrates phosphorylation site motif. Hydrophobic amino acids at positions -3 and -1, and negatively charged amino acids at position +1 are positive determinants for substrate phosphorylation, while positively charged residues surrounding the S/TQ are negative determinants. The complex phenotypes of cells derived from patients with AT suggests that ATM has additional cellular substrates. ATM is present as an inactive homodimer or multimer prior to activation. DNA double-stranded breaks induced by ionizing radiation (IR) cause rapid ATM autophosphorylation at Ser1981 in human or the Ser1987 equivalent in mouse.

Specificity

Reacts with ATM phosphorylated at serine 1981. Clone 10H11.E12 is covered by US patent No. 6,916,627 and 7,108,992.

Immunogen

Epitope: pSer1981.
KLH-conjugated, synthetic peptide corresponding to amino acids 1974-1988 (SLAFEEG[pS]QSTTISS) of human ATM. The immunizing sequence is 11/12 identical to the corresponding mouse and rat sequence.

Application

Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12, Alexa Fluor 488 is an antibody against phospho-ATM (Ser1981) for use in Immunocytochemistry.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
The unconjugated antibody (Cat. Nos. 05-740, MAB3806 & MAB3806-C) is shown to be suitable also for ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, and Western blotting applications. PE-conjugate (Cat. No. FCMAB110P) is also available and tested for Flow cytometry application.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected UV-induced ATM Ser1981 phosphorylation in HeLa cells.

Target description

The unconjugated antibody (Cat. No. 05-430) is shown to recognize the ~370 kDa phosphoATM target band and a >400 kDa non-specific band.

Physical form

Protein G Purified
Purified mouse monoclonal IgG1κ antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service