Skip to Content
Merck
All Photos(4)

Key Documents

05-729

Sigma-Aldrich

Anti-Bcl2 Antibody, clone 100

clone 100, Upstate®, from mouse

Synonym(s):

B-cell CLL/lymphoma 2, B-cell lymphoma protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

100, monoclonal

species reactivity

feline, canine, human

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (Paraffin)
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BCL2(596)

General description

Apoptosis regulator Bcl-2 (UniProt: P10415; also known as Bcl2) is encoded by the BCL2 gene (Gene ID: 596) in human. The members of Bcl-2 family of proteins are known to be involved in either blocking or promoting apoptotic cell death. They act at checkpoints upstream of caspases and share regions of homology termed BH (Bcl Homology) domains. All members of the Bcl-2 family (except Bad and Bid) contain a hydrophobic C-terminus (transmembrane domain), which helps them to attach to membrane. Bcl-2 and other anti-apoptotic members of the Bcl-2 family are located in the outer mitochondrial membrane. Bcl-2 is a single-pass membrane protein that is localized in endoplasmic reticulum, nucleus, and mitochondrial membrane. In mitochondria it is especially enriched at contact sites where the inner and outer membranes come in close proximity. It is reported to counteract the action of pore forming Bax protein. Its transmembrane domain is localized in amino acids 212-233. It contains four BH motifs. BH1 (aa 10-30), BH2 (aa 93-107), and BH4 (aa 136-155) are required for its anti-apoptotic activity. Bcl-2 can be present in cells as homodimer or it can form heterodimers with BAX, BAD, BAK and Bcl-X(L). Heterodimerization with BAX requires intact BH1 and BH2 motifs and is necessary for anti-apoptotic activity. Bcl-2 is shown to suppress apoptosis by controlling the mitochondrial membrane permeability. It functions in a feedback loop system with caspases and is shown to inhibit caspase activity either by preventing the release of cytochrome c from the mitochondria and/or by binding to the apoptosis-activating factor (APAF-1). During apoptosis it is proteolytically cleaved by caspases and the cleaved protein lacking the BH4 motif displays pro-apoptotic activity. Following growth factor stimulation, Bcl-2 can undergo phosphorylation at serine 70 by PKC, which is required for its anti-apoptotic activity.

Specificity

Predicted to cross-react with feline and canine, based on sequence homology.
This antibody recognizes Bcl2, Mr 26 kDa.

Immunogen

Epitope: a.a. 41-54
Synthetic peptide corresponding to amino acids 41-54 of human Bcl2 protein. The immunizing sequence is identical in canine and feline.

Application

Flow Cytometry Analysis: A representative lot of this antibody detected Bcl2 in Jurkat cells.
Western Blotting Analysis: 1μg/mL of this antibody detected Bcl2 in Raji cell lysate.
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected Bcl2 in Human tonsil tissue sections.

Quality

Routinely evaluated by western blot on Raji RIPA cell lysate.

Western Blot Analysis:
0.5-2 µg/mL of this lot detected Bcl2 in a Raji RIPA cell lysate.

Target description

26 kDa

Linkage

Replaces: 04-436

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 in buffer containing 70% storage buffer PBS with 0.1% sodium azide and 30% glycerol. Store at -20ºC.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20ºC. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20ºC may cause glycerol-containing solutions to become frozen during storage.

Analysis Note

Control
Tonsil tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Pezzella et al.
British journal of cancer, 65(1), 87-89 (1992-01-01)
Conflicting results have been published on the prognostic significance of t(14;18) in follicular lymphoma: Yunis et al. (1989) reported that its presence indicated poor response to therapy and short survival, whereas Levine et al. (1988) showed no difference in prognosis
VIP and VIP gene silencing modulation of differentiation marker N-cadherin and cell shape of corneal endothelium in human corneas ex vivo.
Koh, SW; Chandrasekara, K; Abbondandolo, CJ; Coll, TJ; Rutzen, AR
Investigative Ophthalmology & Visual Science null
MART-10, a New Generation of Vitamin D Analog, Is More Potent than 1?,25-Dihydroxyvitamin D(3) in Inhibiting Cell Proliferation and Inducing Apoptosis in ER+ MCF-7 Breast Cancer Cells.
Chiang, KC; Yeh, CN; Chen, SC; Shen, SC; Hsu, JT; Yeh, TS; Pang, JH; Su, LJ; Takano et al.
Evidence-Based Complementary and Alternative Medicine : ECAM null
Lysandra Castro et al.
Expert opinion on environmental biology, 5(Suppl 1) (2016-08-12)
Genistein, an estrogenic, soy-derived isoflavone, may play a protective role against hormone-related cancers. We have reported that a high concentration of genistein inhibits cell proliferation and induces apoptosis in human uterine smooth muscle cells, but not in leiomyoma (fibroid) cells.
Human lactoferrin exerts bi-directional actions on PC12 cell survival via ERK1/2 pathway.
Lin, Tsung-Yao, et al.
Biochemical and biophysical research communications, 337, 330-336 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service