Skip to Content
Merck
All Photos(1)

Key Documents

05-710

Sigma-Aldrich

Anti-NG2 Antibody, clone 132.38

clone 132.38, Upstate®, from mouse

Synonym(s):

Anti-CSPG4A, Anti-HMW-MAA, Anti-MCSP, Anti-MCSPG, Anti-MEL-CSPG, Anti-MSK16, Anti-NG2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

132.38, monoclonal

species reactivity

rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Cspg4(121021)
rat ... Cspg4(81651)

Specificity

Recognizes NG2.

Immunogen

HEK293 cells expressing a truncated integral membrane form of NG2 consisting of amino acids 1592-2222

Application

Anti-NG2 Antibody, clone 132.38 detects level of NG2 & has been published & validated for use in IH & WB.
Cross-reactivity expected with mouse.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Quality

routinely evaluated by immunoblot on whole rat brain preparation

Target description

~270-300 kDa

Physical form

Format: Purified
Protein A Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide as a preservative.
Protein G Purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Levine et al.
Trends in neurosciences, 24(1), 39-47 (2001-02-13)
Adult oligodendrocyte precursor cells (OPCs) make up around 5-8% of the glial cell population in the CNS. Their function in the undamaged CNS is largely unknown, but their processes are in contact with nodes of Ranvier and synapses, suggesting a
Sandra Stelzer et al.
BMC neuroscience, 11, 27-27 (2010-02-27)
Junctional adhesion molecule-A (JAM-A) is an adhesive protein expressed in various cell types. JAM-A localizes to the tight junctions between contacting endothelial and epithelial cells, where it contributes to cell-cell adhesion and to the control of paracellular permeability. So far
In vivo intracellular recording suggests that gray matter astrocytes in mature cerebral cortex and hippocampus are electrophysiologically homogeneous.
Mishima, T; Hirase, H
The Journal of Neuroscience null
Marijana Miljkovic-Licina et al.
Molecular cancer therapeutics, 11(12), 2588-2599 (2012-09-25)
Antiangiogenic drugs have been used as anticancer agents to target tumor endothelial cells or pericytes. Because of limited efficacy of the current monotherapies, there is a strong demand for the dual targeting of endothelial cells and pericytes. Here, we identify
Stuart William Smith et al.
Nephron, 131(4), 265-277 (2015-12-04)
Tissue fibrosis and microvascular rarefaction are hallmarks of progressive renal disease. CD248 is a transmembrane glycoprotein expressed by key effector cells within the stroma of fibrotic kidneys including pericytes, myofibroblasts and stromal fibroblasts. In human disease, increased expression of CD248

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service