Skip to Content
Merck
All Photos(3)

Key Documents

05-1339

Sigma-Aldrich

Anti-Trimethyl Histone H3 (Lys4) Antibody, clone CMA304

clone CMA304, from mouse

Synonym(s):

H3K4me3, Histone H3 (tri methyl K4), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CMA304, monoclonal

species reactivity

human, vertebrates

technique(s)

ELISA: suitable
dot blot: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
multiplexing: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys4)

Gene Information

human ... H3C1(8350)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome. Histones are modified post-translationally; and these modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. The modifications occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Trimethylation of histone H3 (Lys4) is is found at the 5′ end of active genes and contributes to transcriptional activation by recruiting chromatin-remodelling enzymes. Accordingly, the inactive X chromosome is largely devoid of this mark.

Specificity

Broad species cross-reactivity is expected, based on sequence homology.
This antibody specifically recognizes Histone H3 trimethylated at Lys4. The antibody binding specificity allows for modifications of Lys91.

Immunogen

Epitope: Trimethylated Lys4
Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, trimethylated on Lys4, conjugated to KLH.

Application

Anti-Trimethyl Histone H3 (Lys4) Antibody, clone CMA304 is a mouse monoclonal antibody for detection of Trimethyl Histone H3 (Lys4) also known as H3K4me3, Histone H3 (tri methyl K4) & has been validated in ELISA, ICC, IP, WB, Mplex, DB.
ELISA:
This antibody has been shown by an outside laboratory to be suitable for ELISA.1

Immunocytochemistry:
This antibody has been shown by an outside laboratory to be suitable for immunocytochemistry.1

Immunoprecipitation:
This antibody has been shown by an outside laboratory to be suitable for immunoprecipitation.1

Multiplexing:
This antibody specifically recognizes histone H3 trimethylated on Lys4 by Luminex® assay.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Western Blot Analysis:
0.5 - 2 μg/mL (1:500-2000) dilution of this antibody detected trimethyl Histone H3 (Lys4) on 10 μg of HeLa acid extract.

Target description

approx. 17 kDa

Linkage

Replaces: 04-791

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt. For maximum recovery of product, centrifuge the vial prior to removing the cap. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa Acid extract lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Luminex is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Shen et al.
Cell stem cell, 15(4), 459-471 (2014-10-04)
With the exception of imprinted genes and certain repeats, DNA methylation is globally erased during preimplantation development. Recent studies have suggested that Tet3-mediated oxidation of 5-methylcytosine (5mC) and DNA replication-dependent dilution both contribute to global paternal DNA demethylation, but demethylation
Diego Villar et al.
Cell, 160(3), 554-566 (2015-01-31)
The mammalian radiation has corresponded with rapid changes in noncoding regions of the genome, but we lack a comprehensive understanding of regulatory evolution in mammals. Here, we track the evolution of promoters and enhancers active in liver across 20 mammalian
Arginine methylation at histone H3R2 controls deposition of H3K4 trimethylation.
Kirmizis, Antonis, et al.
Nature, 449, 928-932 (2007)
Trimethylation of histone H3 lysine 4 is an epigenetic mark at regions escaping mammalian X inactivation.
Khalil, Ahmad M and Driscoll, Daniel J
Epigenetics, 2, 114-118 (2007)
Idelisse Ortiz Torres et al.
Nature communications, 6, 6204-6204 (2015-02-18)
The retinoblastoma binding protein KDM5A removes methyl marks from lysine 4 of histone H3 (H3K4). Misregulation of KDM5A contributes to the pathogenesis of lung and gastric cancers. In addition to its catalytic jumonji C domain, KDM5A contains three PHD reader

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service