Skip to Content
Merck
All Photos(1)

Key Documents

V-055

Supelco

(-)-Riboflavin-13C4,15N2 (Vitamin B2-13C4,15N2) solution

100 μg/mL (1% ammonium acetate in 50:50 methanol:water), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C1313C4H20N215N2O6
CAS Number:
Molecular Weight:
382.32
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (1% ammonium acetate in 50:50 methanol:water)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

General description

Riboflavin is an essential B vitamin for maintaining human health with key roles in energy metabolism as well as a critical component of enzyme cofactors and flavoproteins.

Application

This stable-labeled internal standard is suitable for quantification of Riboflavin in a variety of testing applications by LC/MS or GC/MS from diagnostic testing of vitamin B2 deficiency to quality control testing of vitamin B2-based dietary supplements, fortified foods or pharmaceutical preparations.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service