Skip to Content
Merck
All Photos(1)

Key Documents

C-111

Supelco

Catecholamine Metabolites Mix solution

1 mg/mL each component in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O5
Molecular Weight:
198.17
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Homovanillic acid
5-Hydroxyindole-3-acetic acid
(±)-Vanilmandelic acid

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL each component in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
forensics and toxicology

format

multi-component solution

storage temp.

−70°C

General description

A Certified Spiking Solution® of major catecholamine metabolites 5-Hydroxyindole-3-acetic acid (5-HIAA), Homovanillic acid (HVA), and (±)-Vanilmandelic acid (VMA). This multi-component mix is suitable for use in preparation of linearity standards, calibrators, or controls as well as for other critical quantitative applications such as internal proficiency testing or for OEM use in preparation of reagents, assay development, and instrument IQ/OQ. Certified concentration values for each component provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples.
.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    DL-4-Hydroxy-3-methoxymandelic acid
    5-Hydroxyindole-3-acetic acid
    Homovanillic acid

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sravanth K Hindupur et al.
Nature, 555(7698), 678-682 (2018-03-22)
Histidine phosphorylation, the so-called hidden phosphoproteome, is a poorly characterized post-translational modification of proteins. Here we describe a role of histidine phosphorylation in tumorigenesis. Proteomic analysis of 12 tumours from an mTOR-driven hepatocellular carcinoma mouse model revealed that NME1 and
Joao Rocha et al.
Basic & clinical pharmacology & toxicology, 116(5), 398-413 (2014-10-08)
Rosmarinic acid is a polyphenolic compound and main constituent of Rosmarinus officinalis and has been shown to possess antioxidant and anti-inflammatory properties. We aimed to evaluate the anti-inflammatory properties of rosmarinic acid and of an extract of R. officinalis in
Adriana Junqueira et al.
Arquivos brasileiros de cardiologia, 106(1), 18-25 (2015-12-10)
Although the beneficial effects of resistance training (RT) on the cardiovascular system are well established, few studies have investigated the effects of the chronic growth hormone (GH) administration on cardiac remodeling during an RT program. To evaluate the effects of
Verena Albert et al.
EMBO molecular medicine, 8(3), 232-246 (2016-01-17)
Activation of non-shivering thermogenesis (NST) in brown adipose tissue (BAT) has been proposed as an anti-obesity treatment. Moreover, cold-induced glucose uptake could normalize blood glucose levels in insulin-resistant patients. It is therefore important to identify novel regulators of NST and
Z K Saritas et al.
Bratislavske lekarske listy, 115(8), 508-513 (2014-09-24)
The study included 12 clinically healthy, adult male dogs of various breeds, admitted to our clinic for castration. After general anesthesia with sevoflurane, we administered epidural fentanyl (1 mcg/kg) to fentanyl group, while lidocaine group was given Lidocaine (3 mg/kg)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service