Skip to Content
Merck
All Photos(2)

Key Documents

850114P

Avanti

IP3(1,3,5)

Avanti Research - A Croda Brand 850114P, powder

Synonym(s):

D-myo-inositol-1,3,5-triphosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H24N3O15P3
CAS Number:
Molecular Weight:
471.19
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850114P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850114P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850114P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]([C@H](O)[C@H](OP(O)([O-])=O)[C@@H](O)[C@@H]1OP([O-])(O)=O)OP(O)([O-])=O.[NH4+].[NH4+].[NH4+]

General description

Inositol 1,4,5-triphosphate (IP3), a secondary messenger can bind to the IP3 receptor. The 1,3,5-inositol trisphosphate is formed due to phosphatidyl inositol breakdown, binds to an intracellular receptor, which enhances intracellular calcium.

Biochem/physiol Actions

The inositol triphosphate (IP3)-signaling pathway may be linked with several progressive and biological processes in plants.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850114P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850114P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular characterization of At5PTase1, an inositol phosphatase capable of terminating inositol trisphosphate signaling
Berdy SE, et al.
Plant Physiology, 126(2), 801-810 (2001)
Receptor signaling and the cell biology of synaptic transmission
Handbook of Clinical Neurophysiology, 106, 9-35 (2012)
Receptor signaling and the cell biology of synaptic transmission
Yu JZ and Rasenick MM
Handbook of Clinical Nutrition and Aging (2002)
Molecular characterization of At5PTase1, an inositol phosphatase capable of terminating inositol trisphosphate signaling
Berdy SE, et al.
Plant Physiology (2001)
Spinocerebellar ataxia type 11.
Handbook of Clinical Neurophysiology (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service