Skip to Content
Merck
All Photos(2)

Key Documents

830842C

Avanti

08:0 PA

1,2-dioctanoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

PA(8:0/8:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O8PNa
CAS Number:
Molecular Weight:
446.45
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (830842C-10mg)
pkg of 1 × 2.5 mL (830842C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 830842C

concentration

10 mg/mL (830842C-10mg)
10 mg/mL (830842C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC)COC(=O)CCCCCCC)O

InChI

1S/C19H37O8P.Na/c1-3-5-7-9-11-13-18(20)25-15-17(16-26-28(22,23)24)27-19(21)14-12-10-8-6-4-2;/h17H,3-16H2,1-2H3,(H2,22,23,24);/q;+1/p-1/t17-;/m1./s1

InChI key

ZNDNSOYDPUTHKQ-UNTBIKODSA-M

General description

Phosphatidic acid (PA) is a diacyl-glycerophospholipid. Two fatty acids and a phosphate group are attached to a glycerol molecule with the help of covalent bond through ester linkage.[1] PA can be synthesized by the activity of the endogenous enzyme phospholipase D. This prime glycerophospholipid is present in the biomembrane.[2] 08:0 PA (1,2-dioctanoyl-sn-glycero-3-phosphate), a short chain PA[3] is a cell-permeable analog of PA.[4]

Application

08:0 PA has been used to induce differentiation in order to study the effects of phosphatidic acids (PAs) on the inhibition of adipogenic differentiation in 3T3-L1 preadipocytes.[3] It may be used as a mammalian target of rapamycin (mTOR) activator in SGC-996 gall bladder carcinoma cells.[5] It may also be used to study its effects on intracellular Ca2+ concentration ([Ca2+]i) in C6 rat glioma and L2071 mouse fibroblast cells.[6]

Biochem/physiol Actions

Phosphatidic acid (PA) serves as a precursor for phosphatidylcholine or phosphatidylserine. It plays a functional role in influencing the membrane curvature and signaling.[2] C8-PA/08:0 PA can block its association with soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNAREs) to prevent the priming activity on isolated vacuoles. It also has the ability to change the proteolytic cleavage profile of Sec18.[4]

Packaging

5 mL Clear Glass Sealed Ampule (830842C-10mg)
5 mL Clear Glass Sealed Ampule (830842C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew L Starr et al.
The Journal of biological chemistry, 294(9), 3100-3116 (2019-01-09)
Eukaryotic cell homeostasis requires transfer of cellular components among organelles and relies on membrane fusion catalyzed by SNARE proteins. Inactive SNARE bundles are reactivated by hexameric N-ethylmaleimide-sensitive factor, vesicle-fusing ATPase (Sec18/NSF)-driven disassembly that enables a new round of membrane fusion.
Jordan M Joy et al.
Nutrition & metabolism, 11, 29-29 (2014-06-25)
The lipid messenger phosphatidic acid (PA) plays a critical role in the stimulation of mTOR signaling. However, the mechanism by which PA stimulates mTOR is currently unknown. Therefore, the purpose of this study was to compare the effects of various
A Yalcin et al.
Oncogene, 29(1), 139-149 (2009-10-27)
Choline is an essential anabolic substrate for the synthesis of phospholipids. Choline kinase phosphorylates choline to phosphocholine that serves as a precursor for the production of phosphatidylcholine, the major phospholipid constituent of membranes and substrate for the synthesis of lipid
Hae-In Song et al.
Scientific reports, 6, 36968-36968 (2016-11-23)
Phospholipase D1 (PLD1) plays a known role in several differentiation processes, but its role in adipogenic differentiation remains unknown. In the present study, we identified PLD1 as a negative regulator of adipogenic differentiation. We showed that PLD activity was downregulated
Young-Ja Chang et al.
Prostaglandins & other lipid mediators, 83(4), 268-276 (2007-05-15)
Phosphatidic acid (PA) increased intracellular Ca(2+) concentration ([Ca(2+)](i)) in C6 rat glioma and L2071 mouse fibroblast cells. Dioleoyl PA (PA, 18:1) was the most efficacious, followed by dipalmitoyl PA (16:0 PA) and dimyristoyl PA (14:0 PA). Lysophosphatidic acid (LPA) also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service