Skip to Content
Merck
All Photos(2)

Key Documents

810122P

Avanti

14:0-06:0 NBD PC

Avanti Research - A Croda Brand 810122P, powder

Synonym(s):

1-myristoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H58N5O11P
CAS Number:
Molecular Weight:
743.83
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810122P-1mg)
pkg of 5 × 1 mg (810122P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810122P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) is the most abundant phospholipid in eukaryotes and prime metabolite of glycerolipid metabolism. It is a vital component of the mucosal layer of the colon. NBD PC is a fluorophore with 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) labeled to the phosphatidylcholine.

Application

14:0-06:0 NBD PC may be used as a fluorescent marker in the preparation of hyaluronan (HA)-phospholipid medium for tribological studies against live cartilage. It might also be used as a substrate for type IV P-type ATPases (P4-ATPases), in the directed evolution approach to examine the molecular mechanisms of substrate backbone discrimination.

Biochem/physiol Actions

7-nitrobenz-2-oxa-1,3-diazol-4-yl phosphatidylcholine (NBD PC) functions as a fluorescent lipid analog, used in biological membranes to study various cellular processes. It plays a vital role as a strong bilayer-forming lipid.

Packaging

5 mL Amber Glass Screw Cap Vial (810122P-1mg)
5 mL Amber Glass Screw Cap Vial (810122P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa Veselack et al.
Lubricants (Basel, Switzerland), 6(1) (2018-03-13)
Pre-clinical testing of hemiarthroplasty devices requires that the tribological conditions present in vivo with live cartilage be closely duplicated. A current limitation in the tribological testing of live cartilage involves the use of cell-culture media as lubricant. to develop and
Bartholomew P Roland et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(31), E4460-E4466 (2016-07-20)
Phospholipid flippases in the type IV P-type ATPase (P4-ATPases) family establish membrane asymmetry and play critical roles in vesicular transport, cell polarity, signal transduction, and neurologic development. All characterized P4-ATPases flip glycerophospholipids across the bilayer to the cytosolic leaflet of
Yi-Chih Lin et al.
Nature communications, 11(1), 230-230 (2020-01-15)
Annexins are abundant cytoplasmic proteins, which bind to membranes that expose negatively charged phospholipids in a Ca2+-dependent manner. During cell injuries, the entry of extracellular Ca2+ activates the annexin membrane-binding ability, subsequently initiating membrane repair processes. However, the mechanistic action of
Luís M S Loura et al.
Biochimica et biophysica acta, 1778(2), 491-501 (2007-11-21)
We present a combined theoretical (molecular dynamics, MD) and experimental (differential scanning calorimetry, DSC) study of the effect of 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) acyl chain-labeled fluorescent phospholipid analogs (C6-NBD-PC and C12-NBD-PC) on 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) bilayers. DSC measurements reveal that <1 mol% of
Wei Zong et al.
Langmuir : the ACS journal of surfaces and colloids, 34(23), 6874-6886 (2018-05-20)
The success of nanoparticulate formulations in drug delivery depends on various aspects including their toxicity, internalization, and intracellular location. Vesicular assemblies consisting of phospholipids and amphiphilic block copolymers are an emerging platform, which combines the benefits from liposomes and polymersomes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service