Skip to Content
Merck
All Photos(2)

Key Documents

810122C

Avanti

14:0-06:0 NBD PC

Avanti Research - A Croda Brand 810122C

Synonym(s):

1-myristoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H58N5O11P
CAS Number:
Molecular Weight:
743.83
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810122C-1mg)
pkg of 5 × 1 mL (810122C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810122C

concentration

1 mg/mL (810122C-1mg)
1 mg/mL (810122C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

1-myristoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphocholine (Acyl 06:0 NBD PC) comprises NBD (7-nitrobenz-2-oxa-1,3-diazol-4-yl) fluorescent probe.

Application

Acyl 06:0 NBD PC may be used:
  • as a supplement in ice-cold synthetic defined (SD) medium for yeast cultures for flow cytometry studies
  • as a fluorescent marker in hyaluronan-phospholipid based medium (HA–phospholipid medium) for culturing cartilage cells and to visualize lipid vesicle formation
  • in the preparation of small unilamellar vesicles (SUVs) and fluorescence-labeled supported lipid bilayer (SLB) for fluorescence recovery after photobleaching (FRAP) studies

Biochem/physiol Actions

Lipids that are labeled with NBD (7-nitrobenz-2-oxa-1,3-diazol-4-yl) are useful in lipid membranes dynamics studies. Acyl 06:0 NBD PC is used as yeast extract–peptone–dextrose (YPD) medium component in yeast uptake studies to decipher its incorporation.

Packaging

5 mL Amber Glass Screw Cap Vial (810122C-1mg)
5 mL Amber Glass Screw Cap Vial (810122C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phospholipid Vesicles in Media for Tribological Studies against Live Cartilage
Veselack T, et al.
Lubrication Science, 6(1), 19-19 (2018)
Directed evolution of a sphingomyelin flippase reveals mechanism of substrate backbone discrimination by a P4-ATPase
Roland BP and Graham TR
Proceedings of the National Academy of Sciences of the USA, 113(31), E4460-E4466 (2016)
Annexin-V stabilizes membrane defects by inducing lipid phase transition
Lin YC, et al.
Nature Communications, 11(1), 1-13 (2020)
Involvement of LEM3/ROS3 in the Uptake of Phosphatidylcholine with Short Acyl Chains in Saccharomyces cerevisiae
Ono Y, et al.
Bioscience, Biotechnology, and Biochemistry, 73(3), 750-752 (2009)
Yi-Chih Lin et al.
Nature communications, 11(1), 230-230 (2020-01-15)
Annexins are abundant cytoplasmic proteins, which bind to membranes that expose negatively charged phospholipids in a Ca2+-dependent manner. During cell injuries, the entry of extracellular Ca2+ activates the annexin membrane-binding ability, subsequently initiating membrane repair processes. However, the mechanistic action of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service