Key Documents
Z680826
Duran® stirred reactor caps and accessories, for GL 80 stirred reactor systems
anchor type magnetic stirrer.
Synonym(s):
1200383
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
packaging
pack of 1 ea
manufacturer/tradename
DURAN® 1200383
diam.
62 mm
General description
These connections fit the GL 80 stirred reactor systems which provide addition or removal of media from the bottle during the mixing process.
  • The accessories can be autoclaved and are suitable for use in the biological sector
  • The agitating elements are exchangeable and use up to 500 rpm is possible
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)