Key Documents
Z177482
Scienceware® gas-dispersion tube
polyethylene
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
polyethylene
packaging
pack of 4 ea
manufacturer/tradename
Bel-Art F13691-0000
General description
Linear bell-mouthed fitting has sealed-in PE disc, 70 μm porosity, at large end; 30.5 cm (12 in.) L × 6.35 mm (1/4 in.) I.D. Tube is fitted to the other end, 2.54 cm bell (1 in.) O.D.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP