WHA5230090
Whatman® filter paper, Grade 230, creped
circles, diam. 90 mm (3.5 in.), pkg of 50 ea
Synonym(s):
Whatman filter, Whatman paper, Z761907, paper filter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
circles
creped white cellulose
packaging
pkg of 50 ea
manufacturer/tradename
Cytiva
Whatman 5230-090
parameter
fast flow rate
fast flow rate
diam.
90 mm (3.5 in.)
thickness
0.40 mm (0.02 in.)
pore size
25-30 μm
basis weight
123 g/m2
capacity
high loading
Looking for similar products? Visit Product Comparison Guide
General description
Filter papers are manufactured primarily for non-critical industrial laboratory filtration applications requiring inexpensive filters of consistent performance.
Grade 230 is a strong, thick creped paper widely used in oil chemistry. Retains coarse particles, has a fast filtration speed and high loading capacity.
Note; These papers should not be used for kjeldahl nitrogen analysis.
Grade 230 is a strong, thick creped paper widely used in oil chemistry. Retains coarse particles, has a fast filtration speed and high loading capacity.
Note; These papers should not be used for kjeldahl nitrogen analysis.
Features and Benefits
- Retains coarse particles
- Highspeedand loading capacity
- Strong, thick filter with crêped surface
Legal Information
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service