Skip to Content
Merck
All Photos(1)

Documents

W320218

Sigma-Aldrich

Methyl 2-pyrrolyl ketone

≥98%, FG

Synonym(s):

2-Acetylpyrrole, Methyl 2-pyrrolyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
FEMA Number:
3202
Beilstein:
1882
EC Number:
Council of Europe no.:
4035
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.047
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥98%

bp

220 °C (lit.)

mp

88-93 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

bread; coumarin; licorice; musty; nutty; walnut

SMILES string

CC(=O)c1ccc[nH]1

InChI

1S/C6H7NO/c1-5(8)6-3-2-4-7-6/h2-4,7H,1H3

InChI key

IGJQUJNPMOYEJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • RIFM fragrance ingredient safety assessment, methyl 2-pyrrolyl ketone, CAS Registry Number 1072-83-9.: This research presents a safety assessment of methyl 2-pyrrolyl ketone for its use in fragrances, highlighting its toxicological profile and regulatory considerations (Api AM, Belsito D, Botelho D, et al., 2024, Api et al., 2024).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Ho Lee et al.
The Journal of organic chemistry, 78(3), 1283-1288 (2013-01-08)
A new synthetic route to indolizines with various substituents on the pyridine moiety was developed by utilizing a facile cycloaromatization of 2-acetylpyrrole derivatives. Without isolation, the resulting intermediates were allowed to react with various electrophiles to afford a range of
Ana Filipa L O M Santos et al.
The journal of physical chemistry. A, 113(15), 3630-3638 (2009-03-27)
A combined experimental and computational study on the thermochemistry of 2- and 3-acetylpyrroles was performed. The enthalpies of combustion and sublimation were measured by static bomb combustion calorimetry and Knudsen effusion mass-loss technique, respectively, and the standard (p(o) = 0.1
G C Yen et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 24(12), 1303-1308 (1986-12-01)
Three 2-substituted pyrroles (2-acetylpyrrole, pyrrole-2-carboxaldehyde and pyrrole-2-carboxylic acid), which are products of the Maillard browning reaction, were reacted with nitrite in buffer solution (pH 3) at 50 degrees C for 24 hr. The reaction mixtures were extracted with methylene chloride
Wen-Yong Liu et al.
Journal of Asian natural products research, 5(3), 159-163 (2003-08-23)
Three pyrrole alkaloids were isolated from Bolbostemma paniculatum. Their structures were elucidated as 4-(2-formyl-5-methoxymethylpyrrol-1-yl)butyric acid methyl ester (1), 2-(2-formyl-5-methoxymethylpyrrol-1-yl)-3-phenylpropionic acid methyl ester (2) and alpha-methyl pyrrole ketone (3) by spectroscopic techniques. Among them, 1 and 2 are new compounds.
Nobuo Ochiai et al.
Journal of chromatography. A, 1421, 103-113 (2015-08-16)
An extension of multi-volatile method (MVM) technology using the combination of a standard dynamic headspace (DHS) configuration, and a modified DHS configuration incorporating an additional vacuum module, was developed for milliliter injection volume of aqueous sample with full sample evaporation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service