Skip to Content
Merck
All Photos(3)

Documents

P38706

Sigma-Aldrich

Phthalazine

98%

Synonym(s):

β-Phenodiazine, 2,3-Benzodiazine, 2,3-Diazanaphthalene, Benzo[d]pyridazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N2
CAS Number:
Molecular Weight:
130.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

bp

189 °C/29 mmHg (lit.)

mp

89-92 °C (lit.)

storage temp.

2-8°C

SMILES string

c1ccc2cnncc2c1

InChI

1S/C8H6N2/c1-2-4-8-6-10-9-5-7(8)3-1/h1-6H

InChI key

LFSXCDWNBUNEEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michał Achmatowicz et al.
The Journal of organic chemistry, 74(2), 795-809 (2008-12-18)
p38 MAP kinase inhibitors have attracted considerable interest as potential agents for the treatment of inflammatory diseases. Herein, we describe a concise and efficient synthesis of inhibitor 1 that is based on a phthalazine scaffold. Highlights of our approach include
Manuel Sánchez-Moreno et al.
The Journal of antimicrobial chemotherapy, 67(2), 387-397 (2011-12-01)
To evaluate the in vitro leishmanicidal activity of imidazole-based (1-4) and pyrazole-based (5-6) benzo[g]phthalazine derivatives against Leishmania infantum and Leishmania braziliensis. The in vitro activity of compounds 1-6 was assayed on extracellular promastigote and axenic amastigote forms, and on intracellular
Fadi M Awadallah et al.
European journal of medicinal chemistry, 52, 14-21 (2012-03-24)
New phthalazine-based vasodilators were synthesized through the chloroacylation of the starting compound 1-hydrazinophthalazine 4 to give the two key intermediates 5 and 7. These intermediates were used to alkylate various cyclic amines to furnish the final compounds 6a-h and 8a-h.
Hyungchul Kim et al.
International journal of pharmaceutics, 377(1-2), 105-111 (2009-05-26)
Investigation of the use of solution NMR spectroscopy to determine the effect of organic solvents on chemical shift changes in bases on addition of acids is reported. This information can be useful in the evaluation of solvents and counterion selection
Manuel Sánchez-Moreno et al.
Journal of medicinal chemistry, 55(22), 9900-9913 (2012-10-10)
A series of new phthalazine derivatives 1-4 containing imidazole rings were prepared. The monoalkylamino substituted derivatives 2 and 4 were more active in vitro against T. cruzi and less toxic against Vero cells than both their disubstituted analogues and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service