Skip to Content
Merck
All Photos(1)

Key Documents

GF34186327

Fluorinated Ethylene Propylene Copolymer

(FEP), monofilament, diam. 0.28 mm, L 10 m

Synonym(s):

FP345925

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

monofilament

manufacturer/tradename

Goodfellow 341-863-27

L

10 m

diam.

0.28 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service