Skip to Content
Merck
All Photos(2)

Key Documents

D39800

Sigma-Aldrich

1,10-Dibromodecane

97%

Synonym(s):

Decamethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)10Br
CAS Number:
Molecular Weight:
300.07
Beilstein:
506156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

crystals

refractive index

n20/D 1.4912 (lit.)

bp

160 °C/15 mmHg (lit.)

mp

25-27 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

SMILES string

BrCCCCCCCCCCBr

InChI

1S/C10H20Br2/c11-9-7-5-3-1-2-4-6-8-10-12/h1-10H2

InChI key

GTQHJCOHNAFHRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1,10-Dibromodecane, also known as Decamethylene dibromide, is an aromatic and aliphatic monomer, often utilized in the polymerization reaction to synthesize high molecular weight polysulfides through polycondensation.

Application

1,10-Dibromodecane is used:
  • as a reactant in the Wurtz-type reaction during the synthesis of polyethylene

  • Construction of pillar[4]arene[1]quinone-1,10-dibromodecane pseudorotaxanes in solution and in the solid state.: This research highlights the application of 1,10-Dibromodecane in constructing complex molecular structures known as pseudorotaxanes. The study focuses on the synthesis and stabilization of these structures both in solution and solid states, providing a foundational technique for developing advanced materials in chemical engineering, pharmaceutical synthesis, and high-performance materials within industrial chemical manufacturing sectors. This breakthrough offers potential pathways for new drug delivery systems and smart materials based on the unique properties of these supramolecular assemblies (Sheng et al., 2020).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruili Zhang et al.
Advanced healthcare materials, 9(14), e2000394-e2000394 (2020-06-17)
The complexity of biological systems poses a great challenge in the development of nanotheranostic agents with enhanced therapeutic efficacies. To systematically overcome a series of barriers during in vivo administration and achieve optimal antitumor activity, nanotheranostic agents that can self-adaptively
Synthesis of polysulfides containing s-triazine rings from 6-substituted amino-1, 3, 5-triazine-2, 4-dithiols and 1, 10-dibromodecane
Yoshiyuki O, et al.
Macromolecular Rapid Communications, 20, 294-298 (1999)
Stefania Aivali et al.
The journal of physical chemistry. B, 124(24), 5079-5090 (2020-05-28)
Conjugation-break flexible spacers in-between π-conjugated segments were utilized herein toward processable perylene diimide (PDI)-based polymers. Aromatic-aliphatic PDI-based polymers were developed via the two-phase polyetherification of a phenol-difunctional PDI monomer and aliphatic dibromides. These polyethers showed excellent solubility and film-forming ability
Chemical structures, properties, and applications of selected crude oil-based and bio-based polymers
Piotr K, et al.
Polymers, 14, 5551-5551 (2022)
Design and Synthesis of Imidazolium-Mediated Tro?ger?s Base-Containing Ionene Polymers for Advanced CO2 Separation Membranes
Irshad k, et al.
ACS Omega, 4, 3439-3448 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service