Key Documents
BAF131610005
SP Bel-Art® disposal bag
width × H × thickness 24 in. (610 mm) × 30 in. (760 mm) × 0.04 mm (1.5 mil), pack of 100 ea, translucent polypropylene
Synonym(s):
bag, disposal bag
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
translucent polypropylene
feature
label (Without Biohazard Warning Label or Sterilization Indicator Patch)
Fits waste can 7.3 or 12 gallon
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art F13161-0005
width × H × thickness
24 in. (610 mm) × 30 in. (760 mm) × 0.04 mm (1.5 mil)
General description
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
Features and Benefits
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
  • Transparent polypropylene; 0.03mm (1.5 mil) thick
  • Pass ASTM
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP