Skip to Content
Merck
All Photos(2)

Key Documents

900606

Sigma-Aldrich

Fmoc-Trp-BODIPY

≥95%

Synonym(s):

Trp-BODIPY

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H39BF2N4O4
Molecular Weight:
748.62
UNSPSC Code:
12352111
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

functional group

Fmoc

Application

This compound is an Fmoc-labeled amino acid of tryptophan attached to a BODIPY dye through a spacer-free C-C linkage that can be used in solid phase peptide synthesis (SPPS). Research presented in Mendive-Tapia et al. tested the incorporation of the fluorescent amino acid into an anti-fungal peptide with two tryptophans by replacing either one or both of the residues. The three peptides showed enhanced fluorescence with increasing hydrophobicity of the environment. The peptides also maintained binding to Aspergillus fumigatus and could be detected by fluorescence imaging. That same publication showed that a cyclic version of the peptide containing a single fluorescent amino acid was more resistant to proteolysis than the linear peptide and still useful for imaging. The amino acid can be incorporated during standard conditions for SPPS and can also tolerate mildly acidic conditions of 1% trifluoroacetic acid allowing use of acid-labile solid supports. This amino acid should enable fluorescence detection of peptides for use in a variety of applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Spacer-free BODIPY fluorogens in antimicrobial peptides for direct imaging of fungal infection in human tissue.
Tapia ML, et al.
Nature Communications, 7, 10940-10940 (2016)

Related Content

Optical imaging has revolutionized our understanding of how biological systems behave at a molecular level. In our group we develop Dynamic Activatable Fluorophores (DYNAFLUORS) as chemical probes to image molecular events associated to inflammation and cancer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service