Skip to Content
Merck
All Photos(2)

Documents

767549

Sigma-Aldrich

Poly(ethylene glycol) diacrylate

average Mn 20,000, PEG average Mn 20,000 (n~450), acrylate, ≤1000 ppm MEHQ as inhibitor

Synonym(s):

PEG diacrylate, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) diacrylate, average Mn 20,000, contains ≤1000 ppm MEHQ as inhibitor

form

solid

mol wt

PEG average Mn 20,000 (n~450)
average Mn 20,000

contains

≤1000 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

mp

60-65 °C

Ω-end

acrylate

α-end

acrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

General description

Poly(ethylene glycol) diacrylate (PGEDA) is used for synthesising highly cross-linked hydrogels which are used as biomaterials in tissue engineering. These hydrogels are formed using non-cytotoxic photo initiators. PEG hydrogels can be easily covalently linked to bioactive proteins and peptides which in turn promote specific cell activity either on the surface or within the hydrogel.

Application

This homobifunctional PEG can be used in hydrogel applications; biocompatibilization; thiol-ene coupling; and other applications using cross-linked PEG networks.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia E Leslie-Barbick et al.
Biomaterials, 32(25), 5782-5789 (2011-05-27)
Microvascularization of tissue engineered constructs was achieved by utilizing a VEGF-mimicking peptide, QK, covalently bound to a poly(ethylene glycol) hydrogel matrix. The 15-amino acid peptide, developed by D'Andrea et al., was modified with a PEG-succinimidyl ester linker on the N-terminus
Pilnam Kim et al.
Lab on a chip, 6(11), 1432-1437 (2006-10-27)
We present a simple and widely applicable method to fabricate micro- and nanochannels comprised entirely of crosslinked polyethylene glycol (PEG) by using UV-assisted irreversible sealing to bond partially crosslinked PEG surfaces. The method developed here can be used to form
Won-Gun Koh et al.
Langmuir : the ACS journal of surfaces and colloids, 18(7), 2459-2462 (2002-06-29)
We present an easy and effective method for the encapsulation of cells inside PEG-based hydrogel microstructures fabricated using photolithography. High-density arrays of three-dimensional microstructures were created on substrates using this method. Mammalian cells were encapsulated in cylindrical hydrogel microstructures of
Ruohong Shi et al.
Small (Weinheim an der Bergstrasse, Germany), 16(37), e2002946-e2002946 (2020-08-11)
Hydrogels with the ability to change shape in response to biochemical stimuli are important for biosensing, smart medicine, drug delivery, and soft robotics. Here, a family of multicomponent DNA polymerization motor gels with different polymer backbones is created, including acrylamide-co-bis-acrylamide

Articles

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

See All

Protocols

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service