Skip to Content
Merck
All Photos(1)

Key Documents

739723

Sigma-Aldrich

Glycocholic-2,2,4,4-d4 acid

98 atom % D, 98% (CP)

Synonym(s):

3α,7α,12α-Trihydroxy-5β-cholan-24-oic-2,2,4,4-d4 acid N-(carboxymethyl)amide, N-(3α,7α,12α-Trihydroxy-24-oxocholan-24-yl)-glycine-2,2,4,4-d4, Cholylglycine-2,2,4,4-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26D4H39NO6
CAS Number:
Molecular Weight:
469.65
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

165-168 °C (lit.)

mass shift

M+4

SMILES string

[H][C@@]12[C@]([C@](CC([2H])([2H])[C@@H](O)C3([2H])[2H])(C)[C@]3([H])C[C@H]2O)([H])C[C@H](O)[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC(NCC(O)=O)=O

InChI

1S/C26H43NO6/c1-14(4-7-22(31)27-13-23(32)33)17-5-6-18-24-19(12-21(30)26(17,18)3)25(2)9-8-16(28)10-15(25)11-20(24)29/h14-21,24,28-30H,4-13H2,1-3H3,(H,27,31)(H,32,33)/t14-,15+,16-,17-,18+,19+,20-,21+,24+,25+,26-/m1/s1/i8D2,10D2

InChI key

RFDAIACWWDREDC-WZQUIQSLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Runbin Sun et al.
Drug metabolism and disposition: the biological fate of chemicals, 49(3), 276-286 (2020-12-31)
Our previous study suggests that berberine (BBR) lowers lipids by modulating bile acids and activating intestinal farnesoid X receptor (FXR). However, to what extent this pathway contributes to the hypoglycemic effect of BBR has not been determined. In this study
Diana M Shih et al.
Journal of lipid research, 56(1), 22-37 (2014-11-08)
We performed silencing and overexpression studies of flavin containing monooxygenase (FMO) 3 in hyperlipidemic mouse models to examine its effects on trimethylamine N-oxide (TMAO) levels and atherosclerosis. Knockdown of hepatic FMO3 in LDL receptor knockout mice using an antisense oligonucleotide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service