All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
solid
mp
153-157 °C (lit.)
functional group
ester
ketone
SMILES string
OC1=Cc2ccccc2OC1=O
InChI
1S/C9H6O3/c10-7-5-6-3-1-2-4-8(6)12-9(7)11/h1-5,10H
InChI key
MJKVTPMWOKAVMS-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Valeria Nori et al.
Beilstein journal of organic chemistry, 16, 1084-1091 (2020-06-20)
Cascade cyclocarbopalladation of the readily available aryl/alkyl-substituted propargylic amides containing an aryl iodide moiety, followed by Suzuki-Miyaura coupling with arylboronic acids, allowed an efficient regio- and stereoselective synthesis of tetrasubstituted 4-methylene-3,4-dihydroisoquinolin-1(2H)-ones. Moreover, cascade cyclocarbopalladation, followed by the reaction with 2-alkynyltrifluoroacetanilides
Paweł Mateusz Nowak et al.
Journal of chromatography. A, 1548, 92-99 (2018-03-22)
The use of migration times and peak areas referred to another sample component - internal standard, brings many benefits in improving reliability of capillary electrophoresis. However, it is quite commonly overlooked that despite relative migration time and peak area ratio
Alberto Valdés et al.
Food research international (Ottawa, Ont.), 137, 109368-109368 (2020-11-26)
Several works have been focused on the extraction of polysaccharides, polyphenols and caffeine from spent coffee grounds (SCG) and their application in food formulations, but the peptide bioactivity from SCG protein hydrolysates has never been addressed. In the present work
William Kirk et al.
Journal of fluorescence, 27(3), 895-919 (2017-02-22)
A novel version of the well-known and commercially successful Green Fluorescent Protein (GFP) variant known as EGFP, with an introduced E222H mutation, was produced in this laboratory. Given the current state of hypotheses about the role of glutamate 222, and
T C Marques et al.
Reproduction in domestic animals = Zuchthygiene, 53(1), 226-236 (2017-12-06)
Effects of adding different concentrations of melatonin (10
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service