All Photos(3)
About This Item
Linear Formula:
NCC6H4NCO
CAS Number:
Molecular Weight:
144.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
mp
101-104 °C (lit.)
functional group
isocyanate
nitrile
storage temp.
2-8°C
SMILES string
O=C=Nc1ccc(cc1)C#N
InChI
1S/C8H4N2O/c9-5-7-1-3-8(4-2-7)10-6-11/h1-4H
InChI key
TVSXDZNUTPLDKY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4-Cyanophenyl isocyanate, also known as 4-isocyanatobenzonitrile, is an aryl isocyanate. It alters the surface of silk sericin in lithium chloride/dimethyl sulfoxide solvent by forming urethane linkages.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Linrong Zhu et al.
ChemMedChem, 8(7), 1104-1116 (2013-05-15)
The worldwide prevalence of diabetes has spurred numerous studies on the development of new antidiabetic medicines. As a result, dipeptidyl peptidase IV (DPP4) has been recognized as a validated target. In our efforts to discover new DPP4 inhibitors, we analyzed the
Hidetoshi Teramoto et al.
Biomacromolecules, 5(4), 1392-1398 (2004-07-13)
This paper reports chemical modification of silk sericin in LiCl/dimethyl sulfoxide (DMSO) solvent with 4-cyanophenyl isocyanate. Sericin is a highly hydrophilic protein secreted by Bombyx mori, serving as a protein glue in a cocoon. LiCl/DMSO was found to be a
Reactivity of Hydroxy and Amino Derivatives of 2-Phenyl-1H-imidazoline and 2-Phenyl-1H-imidazole toward Isocyanates: Synthesis of Appropriate Carbamates and Ureas.
Parik P, et al.
Journal of Heterocyclic Chemistry, 50(4), 903-910 (2013)
Design, synthesis and antiproliferative activities of diaryl urea derivatives bearing N-acylhydrazone moiety.
Zhang B, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 23(8), 915-918 (2012)
Xue-Feng Cheng et al.
Chemistry, an Asian journal, 12(1), 45-51 (2016-10-27)
Herein, two molecules based on urea and thiourea, which differ by only a single atom, were designed, successfully synthesized, and fabricated into resistive random-access memory devices (RRAM). The urea-based molecule showed binary write-once-read-many (WORM) storage behavior, whereas the thiourea-based molecule
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service