Skip to Content
Merck
All Photos(3)

Key Documents

476099

Sigma-Aldrich

4-(Trifluoromethyl)phenyl isothiocyanate

97%

Synonym(s):

1-Isothiocyanato-4-(trifluoromethyl)benzene, 1-Isothiocyanato-4-trifluoromethylbenzene, p-Trifluoromethylphenyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4NCS
CAS Number:
Molecular Weight:
203.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

81 °C/11 mmHg (lit.)

mp

39-43 °C (lit.)

functional group

fluoro
isothiocyanate

SMILES string

FC(F)(F)c1ccc(cc1)N=C=S

InChI

1S/C8H4F3NS/c9-8(10,11)6-1-3-7(4-2-6)12-5-13/h1-4H

InChI key

DQEVDFQAYLIBRD-UHFFFAOYSA-N

Related Categories

General description

4-(Trifluoromethyl)phenyl isothiocyanate, also referred as p-trifluoromethylphenylisothiocyanate, is an isothiocyanate derivative.

Application

4-(Trifluoromethyl)phenyl isothiocyanate may be used in the synthesis of 6-[1-amino-3-(4-trifluoromethylphenyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It may also be used in the synthesis of photoinduced electron transfer (PET) sensors.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thorfinnur Gunnlaugsson et al.
The Journal of organic chemistry, 70(26), 10875-10878 (2005-12-17)
[structure: see text] The synthesis and UV-vis and NMR spectroscopic studies of thiourea-based colorimetric sensors for anions are presented. These sensors can recognize anions through hydrogen binding even in competitive pH-buffered aqueous solutions, giving rise to large color changes that
Emma B Veale et al.
Organic & biomolecular chemistry, 7(17), 3447-3454 (2009-08-14)
The thiourea based 4-amino-1,8-naphthalimide molecules 1-5 were designed as fluorescent anion sensors and their photophysical properties investigated upon recognition of biologically relevant anions such as acetate, dihydrogen phosphate and fluoride in DMSO. Synthesised in a single step from their respective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service